MilliporeSigma
  • Quantitative analysis of boeravinones in the roots of Boerhaavia Diffusa by UPLC/PDA.
Quantitative analysis of boeravinones in the roots of Boerhaavia Diffusa by UPLC/PDA.
Phytochemical analysis : PCA (2014-03-29)
Khemraj Bairwa, Amit Srivastava, Sanjay Madhukar Jachak
ABSTRACT
Boerhaavia diffusa is a perennial herb belonging to Nyctaginaceae. Various classes of chemical constituents such as phenolics (boeravinones), terpenoids and organic acids have been reported in B. diffusa roots. As boeravinones have been proposed as putative active constituents for the anti-cancer, spasmolytic and anti-inflammatory activities exhibited by B. diffusa extracts, it is worthwhile developing and validating an ultra-performance liquid chromatography (UPLC) method for analysis of boeravinones in B. diffusa roots. To develop and validate a simple, accurate, robust and rapid UPLC analytical method for quality control of B. diffusa roots. Samples for analysis were prepared by refluxing powdered root material with methanol for 2 h. The extracts were concentrated, dried and stored at -20°C until their use. A UPLC with photodiode array (PDA) method was developed and validated for the quantification of boeravinones in the roots of B. diffusa. The separation of boeravinones was achieved using a BEH Shield C18 -column (2.1 × 100 mm, 1.7 µm) with gradient elution of methanol and water (0.1% acetic acid), at a flow rate of 0.4 mL/min and detection was carried out at λmax 273 nm. The UPLC method developed showed good linearity (r(2)  ≥ 0.9999), accuracy and precision. The UPLC method developed provided a selective, sensitive and rapid analytical method for the quantification of boeravinones in B. diffusa roots. All the validation parameters were found to be within the permissible limits as per International Conference on Harmonisation guidelines.
Product Description
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%