MilliporeSigma
  • The recently identified modifier of murine metastable epialleles, Rearranged L-Myc Fusion, is involved in maintaining epigenetic marks at CpG island shores and enhancers.
The recently identified modifier of murine metastable epialleles, Rearranged L-Myc Fusion, is involved in maintaining epigenetic marks at CpG island shores and enhancers.
BMC biology (2015-04-11)
Sarah K Harten, Harald Oey, Lauren M Bourke, Vandhana Bharti, Luke Isbel, Lucia Daxinger, Pierre Faou, Neil Robertson, Jacqueline M Matthews, Emma Whitelaw
ABSTRACT
We recently identified a novel protein, Rearranged L-myc fusion (Rlf), that is required for DNA hypomethylation and transcriptional activity at two specific regions of the genome known to be sensitive to epigenetic gene silencing. To identify other loci affected by the absence of Rlf, we have now analysed 12 whole genome bisulphite sequencing datasets across three different embryonic tissues/stages from mice wild-type or null for Rlf. Here we show that the absence of Rlf results in an increase in DNA methylation at thousands of elements involved in transcriptional regulation and many of the changes occur at enhancers and CpG island shores. ChIP-seq for H3K4me1, a mark generally found at regulatory elements, revealed associated changes at many of the regions that are differentially methylated in the Rlf mutants. RNA-seq showed that the numerous effects of the absence of Rlf on the epigenome are associated with relatively subtle effects on the mRNA population. In vitro studies suggest that Rlf's zinc fingers have the capacity to bind DNA and that the protein interacts with other known epigenetic modifiers. This study provides the first evidence that the epigenetic modifier Rlf is involved in the maintenance of DNA methylation at enhancers and CGI shores across the genome.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, purum p.a., for separation (of mineral compounds), ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%