MilliporeSigma
  • In vitro and in vivo evaluations of the performance of an indirubin derivative, formulated in four different self-emulsifying drug delivery systems.
In vitro and in vivo evaluations of the performance of an indirubin derivative, formulated in four different self-emulsifying drug delivery systems.
The Journal of pharmacy and pharmacology (2014-06-26)
Nasim Heshmati, Xinlai Cheng, Else Dapat, Philip Sassene, Gerhard Eisenbrand, Gert Fricker, Anette Müllertz
ABSTRACT
Anticancer indirubins are poorly soluble in water. Here, digestion of four self-emulsifying drug delivery systems (SEDDS) containing E804 (indirubin-3'-oxime 2,3-dihydroxypropyl ether) was compared by dynamic lipolysis and bioavailability studies. Used lipids were either medium-chain or long-chain glycerides. SEDDS E804 were developed. In-vitro lipolysis was carried out at pH 6.5 (37°C) by adding pancreatic lipase (800 U/ml) and controlling by CaCl2 and NaOH addition. E804 content was quantified in the aqueous micellar phase and precipitate using HPLC. Oral bioavailability was determined in rats. Plasma drug content was determined by liquid chromatography (LC)-mass spectrometry. All formulations reserved E804 in the aqueous micellar phase up to 60 min. Precipitation proceeded towards the end of lipolysis up to 45%. Lowest level of precipitation (21%) occurred with long-chain lipids (LC-SEDDS). However, lipolysis was not really discriminative between formulations as the drug mainly stayed in solution. Oral administration of formulations resulted in similar bioavailability of E804 with no significantly different area under the concentration curve. Only medium-chain self-nanoemulsifying drug delivery systems revealed shorter Tmax compared with the other formulations. E804 had a similar performance in four lipid/surfactant systems. All formulations increased the bioavailability of E804 with no significant difference.
Product Description
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Calcium chloride
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol