MilliporeSigma
  • Oxytocin down-regulates mesenteric afferent sensitivity via the enteric OTR/nNOS/NO/KATP pathway in rat.
Oxytocin down-regulates mesenteric afferent sensitivity via the enteric OTR/nNOS/NO/KATP pathway in rat.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-10-28)
J Li, B Xue, T Han, K Huang, L Gong, X Ma, K Liu, S Cui, M Zhang, W Kunze, C Liu
ABSTRACT
Oxytocin plays an analgesic role in modulation of nociception and pain. Most work to date has focused on the central mechanisms of oxytocin analgesia, but little is known about whether peripheral mechanisms are also involved. The mesenteric afferent discharge was recorded in vitro. The expressions of oxytocin receptor (OTR) and neuronal nitric oxide synthase (nNOS) in longitudinal muscle myenteric plexus (LMMP) was identified by immunofluorescence. Oxytocin per se had no effect on the jejunal mesenteric afferent discharge, however, it markedly attenuated the bradykinin- or distention-evoked increase of mesenteric afferent discharge, which was mimiced by the nitric oxide (NO) donor sodium nitroprusside (SNP). Pretreatment of either NOS inhibitor L-NAME or NPLA largely reduced the inhibitory effect of oxytocin on bradykinin-evoked mesenteric afferent discharge. Such effect, to a large extent, was also alleviated by N-and P-type voltage-dependent calcium channel antagonists or KATP blocker glibenclamide. In addition, immunofluorescence studies show strong colocalization of OTR with nNOS in LMMP of the rat jejunum. Oxytocin down-regulates the mesenteric afferent sensitivity through nNOS-NO-KATP pathway. Our findings may reveal a new peripheral mechanism for oxytocin analgesia.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.