MilliporeSigma
  • Inter-instrumental method transfer of chiral capillary electrophoretic methods using robustness test information.
Inter-instrumental method transfer of chiral capillary electrophoretic methods using robustness test information.
Journal of chromatography. A (2014-06-17)
Bart De Cock, Agnieszka Borsuk, Bieke Dejaegher, Johan Stiens, Debby Mangelings, Yvan Vander Heyden
ABSTRACT
Capillary electrophoresis (CE) is an electrodriven separation technique that is often used for the separation of chiral molecules. Advantages of CE are its flexibility, low cost and efficiency. On the other hand, the precision and transfer of CE methods are well-known problems of the technique. Reasons for the more complicated method transfer are the more diverse instrumental differences, such as total capillary lengths and capillary cooling systems; and the higher response variability of CE methods compared to other techniques, such as liquid chromatography (HPLC). Therefore, a larger systematic change in peak resolutions, migration times and peak areas, with a loss of separation and efficiency may be seen when a CE method is transferred to another laboratory or another type of instrument. A swift and successful method transfer is required because development and routine use of analytical methods are usually not performed in the same laboratory and/or on the same type of equipment. The aim of our study was to develop transfer rules to facilitate CE method transfers between different laboratories and instruments. In our case study, three β-blockers were chirally separated and inter-instrumental transfers were performed. The first step of our study was to optimise the precision of the chiral CE method. Next, a robustness test was performed to identify the instrumental and experimental parameters that were most influencing the considered responses. The precision- and the robustness study results were used to adapt instrumental and/or method settings to improve the transfer between different instruments. Finally, the comparison of adapted and non-adapted transfers allowed deriving some rules to facilitate CE method transfers.
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%