MilliporeSigma
  • Efficient differentiation of human embryonic stem cells toward dopaminergic neurons using recombinant LMX1A factor.
Efficient differentiation of human embryonic stem cells toward dopaminergic neurons using recombinant LMX1A factor.
Molecular biotechnology (2014-11-09)
Ali Fathi, Hassan Rasouli, Meghdad Yeganeh, Ghassem Hosseini Salekdeh, Hossein Baharvand
ABSTRACT
Direct differentiation of dopaminergic (DA) neurons from human pluripotent stem cells (hPSCs) in the absence of gene manipulation is the most desired alternative to clinical treatment of Parkinson disease. Protein transduction-based methods could be efficient, safe approaches to enhance direct differentiation of human embryonic stem cells (hESCs) to DA neurons. In the present study, we compared the differentiation efficiency of DA neurons from hESCs with and without the application of LIM homeobox transcription factor 1 alpha (LMX1A), a master regulatory protein in the development of the midbrain neurons and SHH proteins. The results obtained revealed that the treatment of hESCs with recombinant LMX1A (rLMX1A) protein along with dual SMAD inhibition led to higher expression of LMX1B, LMX1A, FOXA2, PITX3, EN1, and WNT1 effector endogenous genes and two-fold expression of PITX3. Moreover, the highest expression level of PITX3 and TH was observed when rLMX1A was added to the induction medium supplemented with SHH. To our best knowledge, this is the first report demonstrating the application of TAT-LMX1A recombinant protein to enhance hESC differentiation to DA as shown by the expression of DA specific makers. These findings pave the way for enhancing the differentiation of hESCs to DA neurons safely and efficiently without genetic modification.
Sigma-Aldrich
Glial Cell Line-derived Neurotrophic Factor human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, analytical standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Millipore
HIS-Select® Nickel Affinity Gel, (1:1 suspension in a 20% ethanol solution)
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade