MilliporeSigma
  • Comparison of chromatographic conditions for analysis of selected psychotropic drugs in human serum.
Comparison of chromatographic conditions for analysis of selected psychotropic drugs in human serum.
Journal of chromatographic science (2014-09-06)
Anna Petruczynik, Karol Wróblewski, Monika Waksmundzka-Hajnos
ABSTRACT
Retention parameters of psychotropic drug standards were determined on different stationary phases: octadecyl silica, polar octadecyl silica, cyanopropyl silica and phenyl-hexyl silica using aqueous eluent systems containing acetonitrile, methanol or mixture of acetonitrile and methanol as organic modifiers; acetic buffer at pH 3.5 and diethylamine. The influence of stationary phases, kind of organic modifier and concentration of methanol and acetonitrile in mobile phases on retention, separation selectivity, peak symmetry and system efficiency was examined. These chromatographic parameters were significantly changed when analyses were performed on different stationary phases, when acetonitrile or methanol was used as organic modifier and when proportions of acetonitrile and methanol in eluent were different. The most efficient and selective systems were applied for quantification of the selected psychotropic drugs in fortified samples of human serum.
Product Description
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis