MilliporeSigma
  • Solvent and pH Effects of Coumarin-Terminated Monolayer on Silver Particles.
Solvent and pH Effects of Coumarin-Terminated Monolayer on Silver Particles.
Journal of fluorescence (2015-06-10)
Issha Nadirah Ismail, Nurul Izzatil Aisya Asri, Hairul Anuar Tajuddin, Zanariah Abdullah
ABSTRACT
A coumarin-terminated self-assembled monolayer on silver particles (C-SAM) from the reduction of silver ions in the presence of compound 3 was successfully prepared by utilizing phase transfer method, and analyzed by FTIR, SEM-EDS, UV-Visible and a particle sizer. The fluorescence behavior of coumarin termini was carried out in ethanol and chloroform with emission wavelength determined at 386 nm, suggesting an interaction between the carbonyl group and the solvent media. The dispersion was then investigated in acidic and basic conditions, showing a direct proportional correlation between the emission and the pH of the aqueous. These results were consistent for interpreting hydrogen bonds, particularly between the carbonyl group with either proton of the alcohol (C=O----H-O-R) or positive species in acidic conditions (C=O----H(+)). The interactions were possible only when the coumarin terminal rearranged in the monolayer and the carbonyl exerted towards the solvent media, while the rest of the molecules were separated from the solvents.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Potassium thioacetate, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
11-Bromo-1-undecanol, 98%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Tetradodecylammonium bromide, ≥99.0% (AT)
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
1,6-Dibromohexane, 96%
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%