MilliporeSigma
  • Elastin organization in pig and cardiovascular disease patients' pericardial resistance arteries.
Elastin organization in pig and cardiovascular disease patients' pericardial resistance arteries.
Journal of vascular research (2015-04-04)
Maria Bloksgaard, Thomas M Leurgans, Inger Nissen, Pia Søndergaard Jensen, Maria Lyck Hansen, Jonathan R Brewer, Luis A Bagatolli, Niels Marcussen, Akhmadjon Irmukhamedov, Lars Melholt Rasmussen, Jo G R De Mey
ABSTRACT
Peripheral vascular resistance is increased in essential hypertension. This involves structural changes of resistance arteries and stiffening of the arterial wall, including remodeling of the extracellular matrix. We hypothesized that biopsies of the human parietal pericardium, obtained during coronary artery bypass grafting or cardiac valve replacement surgeries, can serve as a source of resistance arteries for structural research in cardiovascular disease patients. We applied two-photon excitation fluorescence microscopy to study the parietal pericardium and isolated pericardial resistance arteries with a focus on the collagen and elastin components of the extracellular matrix. Initial findings in pig tissue were confirmed in patient biopsies. The microarchitecture of the internal elastic lamina in both the pig and patient pericardial resistance arteries (studied at a transmural pressure of 100 mm Hg) is fiber like, and no prominent external elastic lamina could be observed. This microarchitecture is very different from that in rat mesenteric arteries frequently used for resistance artery research. In conclusion, we add three-dimensional information on the structure of the extracellular matrix in resistance arteries from cardiovascular disease patients and propose further use of patient pericardial resistance arteries for studies of the human microvasculature.
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Eosin Y Solution, Aqueous