MilliporeSigma
  • Squamasnema amazonica n. gen. n. sp. (Heligmonellinae): A new parasite of Proechimys roberti (Rodentia: Echimyidae) in the Brazilian Amazon.
Squamasnema amazonica n. gen. n. sp. (Heligmonellinae): A new parasite of Proechimys roberti (Rodentia: Echimyidae) in the Brazilian Amazon.
Acta tropica (2015-04-26)
Helrik da Costa Cordeiro, Francisco Tiago de Vasconcelos Melo, Adriano Penha Furtado, Elane Guerreiro Giese, Arnaldo Maldonado, Jeannie Nascimento dos Santos
ABSTRACT
A new species of nematode, Squamasnema amazonica n. gen. n. sp., is described based on specimens found parasitizing the small intestine of Proechimys roberti (Rodentia: Echimyidae) collected during a survey of the fauna of Tapirapé-Aquirí National Forest (Brazil, Eastern Brazilian Amazon). The nematodes were fixed and processed for light microscopy and scanning electron microscopy (SEM). These nematodes were classified under the family Heligmonellidae and the subfamily Heligmonellinae. Although several species in the family Heligmonellidae exhibit discontinuous ridges, Squamasnema n. gen. and Trichotravassosia are the only genera with columns of scales along their entire body, as an apomorphy of the synlophe. Squamasnema n. gen. has columns of cuticular cells along its body, except for on the left flank, and exhibits a synlophe with no size gradient or inclination and does not present chitinized structures supporting the synlophe. Therefore, due to these morphological differences of Squamasnema n. gen., the creation of a new genus was necessary.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.