MilliporeSigma
  • Antivibration gloves: effects on vascular and sensorineural function, an animal model.
Antivibration gloves: effects on vascular and sensorineural function, an animal model.
Journal of toxicology and environmental health. Part A (2015-05-13)
K Krajnak, S Waugh, C Johnson, R G Miller, D Welcome, X Xu, C Warren, S Sarkisian, M Andrew, R G Dong
ABSTRACT
Anti-vibration gloves have been used to block the transmission of vibration from powered hand tools to the user, and to protect users from the negative health consequences associated with exposure to vibration. However, there are conflicting reports as to the efficacy of gloves in protecting workers. The goal of this study was to use a characterized animal model of vibration-induced peripheral vascular and nerve injury to determine whether antivibration materials reduced or inhibited the effects of vibration on these physiological symptoms. Rats were exposed to 4 h of tail vibration at 125 Hz with an acceleration 49 m/s(2). The platform was either bare or covered with antivibrating glove material. Rats were tested for tactile sensitivity to applied pressure before and after vibration exposure. One day following the exposure, ventral tail arteries were assessed for sensitivity to vasodilating and vasoconstricting factors and nerves were examined histologically for early indicators of edema and inflammation. Ventral tail artery responses to an α2C-adrenoreceptor agonist were enhanced in arteries from vibration-exposed rats compared to controls, regardless of whether antivibration materials were used or not. Rats exposed to vibration were also less sensitive to pressure after exposure. These findings are consistent with experimental findings in humans suggesting that antivibration gloves may not provide protection against the adverse health consequences of vibration exposure in all conditions. Additional studies need to be done examining newer antivibration materials.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)