MilliporeSigma
  • Determining the probiotic potential of cholesterol-reducing Lactobacillus and Weissella strains isolated from gherkins (fermented cucumber) and south Indian fermented koozh.
Determining the probiotic potential of cholesterol-reducing Lactobacillus and Weissella strains isolated from gherkins (fermented cucumber) and south Indian fermented koozh.
Research in microbiology (2015-04-04)
Marimuthu Anandharaj, Balayogan Sivasankari, Rajendran Santhanakaruppu, Muthusamy Manimaran, Rizwana Parveen Rani, Subramaniyan Sivakumar
ABSTRACT
This study sought to evaluate the probiotic potential of lactic acid bacteria (LAB) isolated from traditionally fermented south Indian koozh and gherkin (cucumber). A total of 51 LAB strains were isolated, among which four were identified as Lactobacillus spp. and three as Weissella spp. The strains were screened for their probiotic potential. All isolated Lactobacillus and Weissella strains were capable of surviving under low pH and bile salt conditions. GI9 and FKI21 were able to survive at pH 2.0 and 0.50% bile salt for 3 h without losing their viability. All LAB strains exhibited inhibitory activity against tested pathogens and were able to deconjugate bile salt. Higher deconjugation was observed in the presence of sodium glycocholate (P < 0.05). Strain FKI21 showed maximum auto-aggregation (79%) and co-aggregation with Escherichia coli MTCC 1089 (68%). Exopolysaccharide production of LAB strains ranged from 68.39 to 127.12 mg/L (P < 0.05). Moreover, GI9 (58.08 μg/ml) and FKI21 (56.25 μg/ml) exhibited maximum cholesterol reduction with bile salts. 16S rRNA sequencing confirmed GI9 and FKI21 as Lactobacillus crispatus and Weissella koreensis, respectively. This is the first study to report isolation of W. koreensis FKI21 from fermented koozh and demonstrates its cholesterol-reducing potential.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading