MilliporeSigma
  • 1,4-dihydroxyanthraquinone electrochemical sensor based on molecularly imprinted polymer using multi-walled carbon nanotubes and multivariate optimization method.
1,4-dihydroxyanthraquinone electrochemical sensor based on molecularly imprinted polymer using multi-walled carbon nanotubes and multivariate optimization method.
Talanta (2015-12-24)
Azizollah Nezhadali, Samaneh Senobari, Maliheh Mojarrab
ABSTRACT
The present work describes development of a simple and cost-effective electrochemical sensor for determination of 1,4-dihydroxyanthraquinone (1,4-DAQ) based on molecularly imprinted polypyrrole (PPY). The molecularly imprinted polymer (MIP) was electrochemically synthesized onto surface of multi-walled carbon-nanotubes-modified pencil graphite electrode (MWCNs-PGE). A computational approach was employed to select the best functional monomer for rational design of MIP. Based on the computational results, pyrrole (PY) was selected as functional monomer. Plackett-Burman design (PBD) was used for selecting the variables which affected the analytical response (current). After screening, the main factors that affect on the MIP-MWCNTs-PGE response efficiency were also optimized using central composite design (CCD). Under the optimized conditions, calibration curve of the imprinted sensor showed a linear concentration range from 10 nmol L(-1) to 100 µmol L(-1), with the limit of detection (LOD) of 4.15 nmol L(-1). The imprinted sensor showed the advantages of high porous surface structure, ease of preparation, good reproducibility and repeatability, high selectivity and sensitivity. Furthermore, the proposed method was successfully extended for the determination of 1,4-DAQ in serum and plasma real samples.
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, 99.97% trace metals basis