MilliporeSigma
Oxytocin facilitates the extinction of conditioned fear in humans.
Biological psychiatry (2014-12-30)
Monika Eckstein, Benjamin Becker, Dirk Scheele, Claudia Scholz, Katrin Preckel, Thomas E Schlaepfer, Valery Grinevich, Keith M Kendrick, Wolfgang Maier, René Hurlemann
ABSTRACT
Current neurocircuitry models of anxiety disorders posit a lack of inhibitory tone in the amygdala during acquisition of Pavlovian fear responses and deficient encoding of extinction responses in amygdala-medial prefrontal cortex circuits. Competition between these two responses often results in a return of fear, limiting control over anxiety. However, one hypothesis holds that a pharmacologic strategy aimed at reducing amygdala activity while simultaneously augmenting medial prefrontal cortex function could facilitate the extinction of conditioned fear. Key among the endogenous inhibitors of amygdala activity in response to social fear signals is the hypothalamic peptide oxytocin. To address the question whether oxytocin can strengthen Pavlovian extinction beyond its role in controlling social fear, we conducted a functional magnetic resonance imaging experiment with 62 healthy male participants in a randomized, double-blind, parallel-group, placebo-controlled design. Specifically, subjects were exposed to a Pavlovian fear conditioning paradigm before receiving an intranasal dose (24 IU) of synthetic oxytocin or placebo. Oxytocin, when administered intranasally after Pavlovian fear conditioning, was found to increase electrodermal responses and prefrontal cortex signals to conditioned fear in the early phase of extinction and to enhance the decline of skin conductance responses in the late phase of extinction. Oxytocin also evoked an unspecific inhibition of amygdalar responses in both phases. Collectively, our findings identify oxytocin as a differentially acting modulator of neural hubs involved in Pavlovian extinction. This specific profile of oxytocin action may open up new avenues for enhancing extinction-based therapies for anxiety disorders.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)