MilliporeSigma
  • Nanoprobing the acidification process during intracellular uptake and trafficking.
Nanoprobing the acidification process during intracellular uptake and trafficking.
Nanomedicine : nanotechnology, biology, and medicine (2015-05-10)
Simone Lerch, Sandra Ritz, Karina Bley, Claudia Messerschmidt, Clemens K Weiss, Anna Musyanovych, Katharina Landfester, Volker Mailänder
ABSTRACT
Many nanoparticular drug delivery approaches rely on a detailed knowledge of the acidification process during intracellular trafficking of endocytosed nanoparticles (NPs). Therefore we produced a nanoparticular pH sensor composed of the fluorescent pH-sensitive dual wavelength dye carboxy seminaphthorhodafluor-1 (carboxy SNARF-1) coupled to the surface of amino-functionalized polystyrene NPs (SNARF-1-NP). By applying a calibration fit function to confocal laser scanning microscopy (CLSM) images, local pH values were determined. The acidification and ripening process of endo/lysosomal compartments containing nanoparticles was followed over time and was found to progress up to 6h to reach an equilibrium pH distribution (maximum pH5.2 [±0.2]). The SNARF-1-NP localization in endo/lysosomal compartments was confirmed by transmission electron microscopy (TEM) and quantitative co-localization analysis with fluorescent endolysosomal marker Rab-proteins by confocal laser scanning microscopy (CLSM). The herein described nanoparticular pH-sensor is a versatile tool to monitor dynamic pH processes inside the endolysosomal compartments. In this interesting article, the authors elegantly designed a nanoparticular pH sensor with fluorescence probe with the capability to measure intracellular and intravesicular pH changes. The application of this method would enable the further understanding of nanoparticle uptake and intracellular physiology.
Product Description
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Aminoethyl methacrylate hydrochloride, contains ~500 ppm phenothiazine as stabilizer, 90%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Fluorescamine, ≥98.0%
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Hexylamine, 99%