MilliporeSigma
  • Combined use of ion mobility and collision-induced dissociation to investigate the opening of disulfide bridges by electron-transfer dissociation in peptides bearing two disulfide bonds.
Combined use of ion mobility and collision-induced dissociation to investigate the opening of disulfide bridges by electron-transfer dissociation in peptides bearing two disulfide bonds.
Analytical chemistry (2015-04-29)
Philippe Massonnet, Gregory Upert, Nicolas Smargiasso, Nicolas Gilles, Loïc Quinton, Edwin De Pauw
ABSTRACT
Disulfide bonds are post-translational modifications (PTMs) often found in peptides and proteins. They increase their stability toward enzymatic degradations and provide the structure and (consequently) the activity of such folded proteins. The characterization of disulfide patterns, i.e., the cysteine connectivity, is crucial to achieve a global picture of the active conformation of the protein of interest. Electron-transfer dissociation (ETD) constitutes a valuable tool to cleave the disulfide bonds in the gas phase, avoiding chemical reduction/alkylation in solution. To characterize the cysteine pairing, the present work proposes (i) to reduce by ETD one of the two disulfide bridges of model peptides, resulting in the opening of the cyclic structures, (ii) to separate the generated species by ion mobility, and (iii) to characterize the species using collision-induced dissociation (CID). Results of this strategy applied to several peptides show different behaviors depending on the connectivity. The loss of SH· radical species, observed for all the peptides, confirms the cleavage of the disulfides during the ETD process.
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
DL-Cysteine, technical grade