MilliporeSigma
  • Evidence for microRNA-31 dependent Bim-Bax interaction preceding mitochondrial Bax translocation during radiation-induced apoptosis.
Evidence for microRNA-31 dependent Bim-Bax interaction preceding mitochondrial Bax translocation during radiation-induced apoptosis.
Scientific reports (2015-10-31)
Ashish Kumar, Soma Ghosh, Sudhir Chandna
ABSTRACT
Downregulation of microRNA-31 has been linked with enhanced stress resistance, while its overexpression leads to cell death. In this study, we found mediatory role of microRNA-31 in γ-radiation-induced apoptosis in a model insect cell line Sf9 carrying well-conserved apoptotic machinery. Mature microRNA-31 is perfectly conserved amongst insects; hence we used biotinylated probes designed from Bombyx mori sequence for its successful detection in Sf9 cells. Target identification using Bombyx mori 3'UTRs predicted miR-31's potential role in Lepidopteran apoptosis, which prompted us to investigate alterations in its expression during radiation-induced cell death. We found significant overexpression of Sf-miR-31 following lethal dose (1,000Gy-3,000Gy) irradiation. Its mediatory role was finally confirmed as antisense-microRNA-31 could successfully inhibit radiation-induced cytochrome-c release, caspase-3 activation and apoptosis. While Bax/Bcl-2 expression remained unchanged, lethal radiation doses induced Bim overexpression and direct Bim-Bax interaction (co-immunoprecipitation) which is not yet unequivocally demonstrated during apoptosis. Quite important, these events were found to be dependent on radiation-induced miR-31 overexpression, as antisense-miR-31 inhibited both the responses and resulted in significant inhibition of cell death. Pro-apoptotic role of miR-31 was further confirmed when miR-31 mimic induced apoptosis involving similar Bim/Bax alterations. Therefore, our study reveals an important mediatory role of miR-31 in radiation-induced cell death.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
HEPES
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
HEPES