MilliporeSigma
  • A Simple Method to Replace Islet Equivalents for Volume Quantification of Human Islets.
A Simple Method to Replace Islet Equivalents for Volume Quantification of Human Islets.
Cell transplantation (2014-05-20)
Karthik Ramachandran, Han-Hung Huang, Lisa Stehno-Bittel
ABSTRACT
Human islets come in a variety of sizes and shapes, and the total volume of islets used for research or clinical transplants must be estimated in a manner that is simple and valid. Islet equivalent (IEQ) measurements are the standard estimate of islet volume. We published a new method (the Kansas method) for estimating rat islet volume using cell numbers that was reliable and valid. Here we modified the method for human islets. We measured the dimensions of isolated human islets showing that they are not spherical and became less so in larger islets, with an average smallest/largest diameter ratio of 0.73 in large islets and 0.85 in small islets. Human islets were individually loaded into 96-well plates, dissociated into single cells, and the total cell number per islet determined with computer-assisted cytometry. Based on the counted cell number per islet, a regression model was created to convert islet diameter to cell number with a high R(2) value (0.99). Separate regression equations for male and female donors or young and old donors were not significantly different than the pooled data and did not improve the regression values. There was an inverse correlation between the cell number per IEQ and islet size. The Kansas method was validated with ATP/cell and cell viability data. Compared to the actual cell count, conventional IEQ measurements overestimated tissue volume of large islets by nearly double. Examples of differences in results obtained from the same data sets normalized to IEQ or the Kansas method included viability and insulin secretion concentrations. The implications of the error associated with the current IEQ method of volume estimation are discussed.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder