MilliporeSigma
  • Study on the interaction of plasma protein binding rate between edaravone and taurine in human plasma based on HPLC analysis coupled with ultrafiltration technique.
Study on the interaction of plasma protein binding rate between edaravone and taurine in human plasma based on HPLC analysis coupled with ultrafiltration technique.
Biomedical chromatography : BMC (2014-12-30)
Dao-quan Tang, Yin-jie Li, Zheng Li, Ting-ting Bian, Kai Chen, Xiao-xiao Zheng, Yan-yan Yu, Shui-shi Jiang
ABSTRACT
In this work, two high-performance liquid chromatography (HPLC) assays were developed and validated for the independent determination of edaravone and taurine using 3-methyl-1-p-tolyl-5-pyrazolone and L-glutamine as internal standards. In in vitro experiments, human plasma was separately spiked with a mixture of edaravone and taurine, edaravone or taurine alone. Plasma was precipitated with acetonitrile containing 0.1% formic acid. Ultrafiltration was employed to obtain the unbound ingredients of the two drugs. The factors that might influence the ultrafiltration effiency were elaborately optimized. Plasma supernatant and ultrafiltrate containing taurine were derivated with o-phthalaldehyde and ethanethiol in the presence of 40 mmol/L sodium borate buffer (pH 10.2) at room temperature within 1 min. Chromatographic separations were achieved on an InertSustain C18 column (250 × 4.6 mm, 5 µm). Isocratic 50 mmol/L ammonium acetate-acetonitrile and gradient 50 mmol/L sodium acetate (pH 5.3)-methanol were respectively selected as the mobile phase for the determination of edaravone and taurine. All of the validation data including linearity, extraction recovery, precision, accuracy and stability conformed to the requirements. Results showed that there were no significant alterations in the plasma protein binding rate of taurine and edaravone, implying that the proposed combination therapy was pharmacologically feasible.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
SAFC
Taurine
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent