Merck
  • Dicyanovinyl heterotetracenes: synthesis, solid-state structures, and photophysical properties.
Dicyanovinyl heterotetracenes: synthesis, solid-state structures, and photophysical properties.
The Journal of organic chemistry (2009-08-29)
Chunyan Du, Jianming Chen, Yunlong Guo, Kun Lu, Shanghui Ye, Jian Zheng, Yunqi Liu, Zhigang Shuai, Gui Yu
ABSTRACT
Two asymmetrically substituted dicyanovinyl heterotetracenes are synthesized and characterized. Single-crystal X-ray diffractions indicate both intermolecular hydrogen bonds and pi-stacking existing in the solid-state structures. The solvatochromic behaviors of the two heterotetracenes were investigated in a variety of solvents. Both experimental and computational results suggest that the two heterotetracenes have low-lying lowest unoccupied molecular orbitals and thus may be potential n-type or bipolar organic semiconductors for organic electronics.
Sigma-Aldrich
4-Bromo-3-nitrobenzaldehyde, 97%