MilliporeSigma
  • Aneuploidy causes premature differentiation of neural and intestinal stem cells.
Aneuploidy causes premature differentiation of neural and intestinal stem cells.
Nature communications (2015-11-18)
Delphine Gogendeau, Katarzyna Siudeja, Davide Gambarotto, Carole Pennetier, Allison J Bardin, Renata Basto
ABSTRACT
Aneuploidy is associated with a variety of diseases such as cancer and microcephaly. Although many studies have addressed the consequences of a non-euploid genome in cells, little is known about their overall consequences in tissue and organism development. Here we use two different mutant conditions to address the consequences of aneuploidy during tissue development and homeostasis in Drosophila. We show that aneuploidy causes brain size reduction due to a decrease in the number of proliferative neural stem cells (NSCs), but not through apoptosis. Instead, aneuploid NSCs present an extended G1 phase, which leads to cell cycle exit and premature differentiation. Moreover, we show that this response to aneuploidy is also present in adult intestinal stem cells but not in the wing disc. Our work highlights a neural and intestine stem cell-specific response to aneuploidy, which prevents their proliferation and expansion.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid