MilliporeSigma
  • Enhanced expression of CD44 variants in human atheroma and abdominal aortic aneurysm: possible role for a feedback loop in endothelial cells.
Enhanced expression of CD44 variants in human atheroma and abdominal aortic aneurysm: possible role for a feedback loop in endothelial cells.
The American journal of pathology (2004-10-29)
Alexandra Krettek, Galina K Sukhova, Uwe Schönbeck, Peter Libby
ABSTRACT
CD44, a polymorphic hyaluronate receptor, may participate in chronic inflammation. We hypothesized that CD44 variants contribute to the development of arterial diseases. CD44 levels vary in normal and diseased arterial tissues in the following order: unaffected arteries < fibrous plaques < or = abdominal aortic aneurysm < atheromatous plaques; and correlate with macrophage content. Furthermore, plaque microvessels express CD44, and anti-CD44v3 or anti-CD44v6 treatment reduces endothelial cell proliferation but not apoptosis in vitro, suggesting functionality of these receptors. Endothelial cells express CD44H and CD44v6 after exposure to interleukin-1beta and tumor necrosis factor-alpha. Macrophages, a major source of abundant CD44 in vitro, express not only CD44H but also variants CD44v4/5, CD44v6, and CD44v7/8, isoforms distinctively regulated by proinflammatory cytokines. Several proinflammatory cytokines induce shedding of CD44 from the surface of macrophages and endothelial cells. Soluble CD44 stimulates the expression and release of interleukin-1beta from endothelial cells, suggesting a positive feedback loop of this cytokine. By demonstrating augmented expression of CD44 and variants within human atheroma and in abdominal aortic aneurysm as well as the vascular cell release of sCD44, a process regulated by proinflammatory cytokines, this study provides new insights on the functions of CD44 in arterial diseases.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution