MilliporeSigma
  • Synthesis, photodynamic activity, and quantitative structure-activity relationship modelling of a series of BODIPYs.
Synthesis, photodynamic activity, and quantitative structure-activity relationship modelling of a series of BODIPYs.
Journal of photochemistry and photobiology. B, Biology (2017-01-21)
Enrico Caruso, Marzia Gariboldi, Alessandro Sangion, Paola Gramatica, Stefano Banfi
ABSTRACT
Here we report the synthesis of eleven new BODIPYs (14-24) characterized by the presence of an aromatic ring on the 8 (meso) position and of iodine atoms on the pyrrolic 2,6 positions. These molecules, together with twelve BODIPYs already reported by us (1-12), represent a large panel of BODIPYs showing different atoms or groups as substituent of the aromatic moiety. Two physico-chemical features (
Sigma-Aldrich
2,4-Dimethylpyrrole, 97%