MilliporeSigma
  • Influence of extreme concentrations of hydrophilic pore-former on reinforced polyethersulfone ultrafiltration membranes for reduction of humic acid fouling.
Influence of extreme concentrations of hydrophilic pore-former on reinforced polyethersulfone ultrafiltration membranes for reduction of humic acid fouling.
Chemosphere (2017-04-04)
Moon Son, Hayoung Kim, Junhyeok Jung, Sungsoo Jo, Heechul Choi
ABSTRACT
To address the issue of membrane fouling by ubiquitous humic substances, a hydrophilic pore-former-blended polyethersulfone UF membrane was successfully synthesized via the phase inversion method. For the first time, extremely high concentrations of polyvinylpyrrolidone (PVP), up to 20 wt%, were tested as the hydrophilic pore-former in order to determine the optimum concentration for humic acid fouling. Intrinsic membrane parameters such as permeability and selectivity were evaluated using a cross-flow UF filtration setup. Interestingly, as little as 1 wt% added PVP can significantly improve membrane permeability. That tiny amount of added PVP increased membrane flux to 1107 L/m
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%