MilliporeSigma
  • Selective precolumn derivatization of fatty acids with the fluorescent tag 6-aminoquinoline and their determination in some food samples by reversed-phase chromatography.
Selective precolumn derivatization of fatty acids with the fluorescent tag 6-aminoquinoline and their determination in some food samples by reversed-phase chromatography.
Electrophoresis (2017-01-29)
Murthy Jonnada, Guadalupe Davila El Rassi, Ziad El Rassi
ABSTRACT
Fatty acids (FAs) have been selectively derivatized with a fluorescent tag, 6-aminoquinoline (6AQ), which yielded fluorescent FA-6AQ derivatives that have excitation (λ
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)