MilliporeSigma
  • Pectin, Hemicellulose, or Lignin? Impact of the Biowaste Source on the Performance of Hard Carbons for Sodium-Ion Batteries.
Pectin, Hemicellulose, or Lignin? Impact of the Biowaste Source on the Performance of Hard Carbons for Sodium-Ion Batteries.
ChemSusChem (2017-04-21)
Xinwei Dou, Ivana Hasa, Maral Hekmatfar, Thomas Diemant, R Jürgen Behm, Daniel Buchholz, Stefano Passerini
ABSTRACT
Hard carbons are currently the most widely used negative electrode materials in Na-ion batteries. This is due to their promising electrochemical performance with capacities of 200-300 mAh g
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%