MilliporeSigma
  • Surface water mitigates the anti-metamorphic effects of elevated perchlorate concentrations in New Mexico spadefoot toad larvae (Spea multiplicata).
Surface water mitigates the anti-metamorphic effects of elevated perchlorate concentrations in New Mexico spadefoot toad larvae (Spea multiplicata).
Environmental science and pollution research international (2017-06-14)
Melissa A Sandoz, Mike Wages, Kim J Wooten, Sheree L Clendening, Lucas R Smith, Brian Mulhearn, Philip N Smith
ABSTRACT
Perchlorate (ClO
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%