MilliporeSigma
  • Expression of a Chimeric Gene Encoding Insecticidal Crystal Protein Cry1Aabc of
Expression of a Chimeric Gene Encoding Insecticidal Crystal Protein Cry1Aabc of
Frontiers in plant science (2017-09-06)
Alok Das, Subhojit Datta, Shallu Thakur, Alok Shukla, Jamal Ansari, G K Sujayanand, Sushil K Chaturvedi, P A Kumar, N P Singh
ABSTRACT
Domain swapping and generation of chimeric insecticidal crystal protein is an emerging area of insect pest management. The lepidopteran insect pest, gram pod borer (
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid