MilliporeSigma
  • Bacterial Expression of a Single-Chain Variable Fragment (scFv) Antibody against Ganoderic Acid A: A Cost-Effective Approach for Quantitative Analysis Using the scFv-Based Enzyme-Linked Immunosorbent Assay.
Bacterial Expression of a Single-Chain Variable Fragment (scFv) Antibody against Ganoderic Acid A: A Cost-Effective Approach for Quantitative Analysis Using the scFv-Based Enzyme-Linked Immunosorbent Assay.
Biological & pharmaceutical bulletin (2017-10-03)
Gorawit Yusakul, Poomraphie Nuntawong, Seiichi Sakamoto, Pahweenvaj Ratnatilaka Na Bhuket, Toshitaka Kohno, Nao Kikkawa, Pornchai Rojsitthisak, Kuniyoshi Shimizu, Hiroyuki Tanaka, Satoshi Morimoto
ABSTRACT
Due to the highly specific binding between an antibody and its target, superior analytical performances was obtained by immunoassays for phytochemical analysis over conventional chromatographic techniques. Here, we describe a simple method for producing a functional single-chain variable fragment (scFv) antibody against ganoderic acid A (GAA), a pharmacologically active metabolite from Ganoderma lingzhi. The Escherichia coli BL21(DE3) strain produced a large amount of anti-GAA scFv. However, in vitro refolding steps, which partially recovered the reactivity of the scFv, were required. Interestingly, the functional scFv was expressed as a soluble and active form in the cytoplasm of an engineered E. coli SHuffle
Sigma-Aldrich
Human TPO / Thyroid Peroxidase ELISA Kit, for serum, plasma, cell culture supernatants and urine