MilliporeSigma
  • Biosorption-based dispersive liquid-liquid microextraction combined with polypyrrole-coated magnetic nanoparticles as an effective sorbent for the extraction of ibuprofen from water samples using magnetic solid-phase extraction.
Biosorption-based dispersive liquid-liquid microextraction combined with polypyrrole-coated magnetic nanoparticles as an effective sorbent for the extraction of ibuprofen from water samples using magnetic solid-phase extraction.
Electrophoresis (2017-07-30)
Seyed Ammar Haeri, Shahryar Abbasi, Sami Sajjadifar
ABSTRACT
In this paper, biosorption-based dispersive liquid-liquid microextraction (BioDLLME) in combination with magnetic solid-phase extraction (MSPE) has been developed as a sample pretreatment method with high enrichment factor for the sensitive determination of ibuprofen in water samples. At first, magnetic Fe
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%