MilliporeSigma
  • 5'-triphosphate-dependent activation of PKR by RNAs with short stem-loops.
5'-triphosphate-dependent activation of PKR by RNAs with short stem-loops.
Science (New York, N.Y.) (2007-12-01)
Subba Rao Nallagatla, Jungwook Hwang, Rebecca Toroney, Xiaofeng Zheng, Craig E Cameron, Philip C Bevilacqua
ABSTRACT
Molecular patterns in pathogenic RNAs can be recognized by the innate immune system, and a component of this response is the interferon-induced enzyme RNA-activated protein kinase (PKR). The major activators of PKR have been proposed to be long double-stranded RNAs. We report that RNAs with very limited secondary structures activate PKR in a 5'-triphosphate-dependent fashion in vitro and in vivo. Activation of PKR by 5'-triphosphate RNA is independent of RIG-I and is enhanced by treatment with type 1 interferon (IFN-alpha). Surveillance of molecular features at the 5' end of transcripts by PKR presents a means of allowing pathogenic RNA to be distinguished from self-RNA. The evidence presented here suggests that this form of RNA-based discrimination may be a critical step in mounting an early immune response.