MilliporeSigma
GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59
GC Analysis of a 12-Component Sulfur Compound Mix on SLB®-IL59 suitable for GC
Materials
related product
Product No.
Description
Pricing
2-Propanethiol
≥95%
Ethyl methyl sulfide
96%
SLB®-IL59 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
Propyl mercaptan
analytical standard
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
oven
30 °C (4 min), 10 °C/min to 300 °C
inj. temp.
250 °C
detector
MSD, interface at 275 °C
carrier gas
helium, 30 cm/sec constant
injection
0.2 μL split 100:1
liner
4 mm I.D., split/splitless type, dual taper design
sample
12 sulfur compounds, neat mix
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany