MilliporeSigma
GC Analysis of Cresols on SLB®-IL60i
GC Analysis of Cresols on SLB®-IL60i suitable for GC
Materials
related product
Product No.
Description
Pricing
p-Cresol
analytical standard
m-Cresol
analytical standard
o-Cresol
analytical standard
SLB®-IL60i Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
used together
Product No.
Description
Pricing
Dichloromethane
HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
CONDITIONS
column
SLB-IL60i, 30 m x 0.25 mm I.D., 0.20 μm (29832-U)
oven
160 °C
inj. temp.
280 °C
detector
MSD, m/z 50-500
MSD interface
260 °C
carrier gas
helium, 27 cm/sec, constant
injection
1 μL 100:1 split
liner
3.5 mm I.D., split/splitless type, wool packed
sample
3 cresols, each at 100 μg/mL in dichloromethane
Description
Analysis Note
Cresols are an important class of industrial synthetic precursors for small molecule synthesis. This application demonstrates the suitability of SLB®-IL60i for the analysis of phenolic compounds, particularly cresols, or methylphenols. The polarity range of the stationary phase provides good selectivity and resolution of these compounds when run isothermally. The inert nature of the i-series column chemistry also provides good peak shape for compounds that tend to tail based on secondary interactions for tradional PMDS and some wax based phases.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany