MilliporeSigma
GC Analysis of FAMEs in Cod Liver Oil on SUPELCOWAX 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of FAMEs in Cod Liver Oil on SUPELCOWAX 10 (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
CONDITIONS
column
SUPELCOWAX 10, 10 m x 0.10 mm I.D., 0.10 μm (25026-U)
oven
180 °C, 40 °C/min to 270 °C (0.5 min)
inj. temp.
280 °C
detector
FID, 280 °C
carrier gas
hydrogen, 100 cm/sec constant
injection
0.2 μL in the split mode (200:1)
sample
cod liver oil FAMEs in hexane
Description
Legal Information
null