MilliporeSigma
GC Analysis of Fragrance Compounds on SUPELCOWAX 10
GC Analysis of Fragrance Compounds on SUPELCOWAX™ 10 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
50 °C (2 min), 2 °C/min to 280 °C (20 min)
inj. temp.
0.2 μL split (100:1)
detector
FID
carrier gas
helium, 25 cm/sec @ 200 °C
Description
Legal Information
SUPELCOWAX is a trademark of Sigma-Aldrich Co. LLC