MilliporeSigma
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH
GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Petrocol® DH Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.50 μm
CONDITIONS
column
Petrocol DH, 100m × 0.25 mm I.D., 0.5 μm film (24160-U)
inj. temp.
250 °C
column temp.
35 °C for 15 min, then to 200 °C at 2 °C/min, hold for 5 min
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec (set at 35 °C)
injection
1 μL
sample
premium unleaded gasoline, split 100:1,
Description
Legal Information
Petrocol is a registered trademark of Merck KGaA, Darmstadt, Germany