MilliporeSigma
GC Analysis of Scotch Whiskey on a Packed Column (5% Carbowax® 20M on 80/120 Carbopack B)
GC Analysis of Scotch Whiskey on a Packed Column (5% Carbowax® 20M on 80/120 Carbopack B) suitable for GC
CONDITIONS
column
2 m × 2 mm I.D. glass
packing
5% Carbowax 20M on 80/120 Carbopack B
oven
70 °C, 4°C/min. to 150 °C
detector
FID
carrier gas
nitrogen, 20 mL/min
injection
1 μL
sample
Scotch Whiskey
Description
Legal Information
Carbowax is a registered trademark of Union Carbide