MilliporeSigma
HPLC Analysis of Lorazepam Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Lorazepam Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materials
related product
Product No.
Description
Pricing
(±)-Lorazepam
Astec® CHIROBIOTIC® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
methanol
flow rate
2 mL/min
column temp.
23 °C
detector
UV, 220 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC