MilliporeSigma
HPLC Analysis of Polar Neutral and Acidic Amino Acids on Ascentis® Si (Silica)
HPLC Analysis of Polar Neutral and Acidic Amino Acids on Ascentis® Si (Silica) application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Si, 10 cm × 2.1 mm I.D., 5 μm particles (581508-U)
mobile phase
[A] 100 mM ammonium formate, pH 3.0, with formic acid; [B] water, [C] acetonitrile
gradient
5% A constant; 10% B, 85% C to 16% B, 79% C in 20 min
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI (+), full scan
injection
2 μL
sample
100 μg/mL in 10:90, (50 mM ammonium formate / formic acid, pH 3.0):acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany