MilliporeSigma
UHPLC-MS Analysis of Basic Drugs and Metabolites with Titan C18
UHPLC-MS Analysis of Basic Drugs and Metabolites with Titan C18 application for UHPLC-MS
Materials
Related product
Product No.
Description
Pricing
Haloperidol metabolite II
analytical standard
Haloperidol
powder
Fluoxetine hydrochloride
solid
CONDITIONS
column
Titan C18 10 cm x 2.1 mm, 1.9 μm particles (577124-U)
mobile phase
[A] Water; [B] acetonitrile, both with 0.1% formic acid
gradient
35%B Isocratic
flow rate
0.4 mL/min
pressure
484 psi (7120)
column temp.
25 °C
detector
MS, ESI(+), MRM
injection
1 μL
sample
50 ng/mL in 50% methanol
Description
Other Notes
MRM transitions
1. Quinidine325.2/81.2
2. Propranolol260.1/116.2
3. Haloperidol376.1/165.1
4. Haloperidol metabolite II378.1/149.1
5. Haloperidol cholor analog392.1/181.0
6. Fluoxetine 310.1/44.2
Legal Information
null