MilliporeSigma
HomePhotometry & ReflectometryPhotometric Determination of Aluminum in Soil Using Chrome Azurole S
Photometric determination of aluminum in soil using chrome azurole S (CAS) subsequent to acid extraction
three models of Spectroquant® spectrophotometers: Prove 100 plus, Prove 300 plus, and Prove 600 plus. Each device has a sleek, modern design with a rectangular body. The largest model in the foreground, Prove 600 plus, is green with a black top and features a touchscreen interface displaying measurement data, as well as a slot for sample insertion. The top right corner of the device has a yellow "M" logo and "600+" printed in bold yellow on the front corner. Two smaller models, Prove 100 plus and Prove 300 plus, are placed in the background. Prove 100 plus is light green with a black top, while Prove 300 plus is yellow with a black top. Both also feature touchscreens and the "M" logo.
Introduction
The assessment of mobile aluminum in soils is essential for evaluating soil quality and its suitability for plant growth, particularly in acidic environments where Al³⁺ toxicity can limit nutrient uptake and impair crop productivity.1 Elevated aluminum levels can interfere with the availability of key nutrients such as phosphorus, calcium, and magnesium, making its routine monitoring important for agricultural and environmental analyses.1
Photometric techniques are commonly used for the routine determination of aluminum in soil extracts due to their simplicity and suitability for laboratory workflows. As Al³⁺ ions do not exhibit intrinsic absorbance in the visible region, these methods rely on the formation of colored complexes between aluminum ions and suitable chromogenic reagents, enabling quantification in the visible range.1
Experimental
This application note details the photometric determination of aluminum in soils, using the Chromazurole S method, after its extraction with acid solution. 
Method
In weakly acidic, acetate-buffered solution aluminum ions react with chrome azurole S to form a blue-violet compound that is determined photometrically. Due to the intrinsic orange coloration of the reagent blank, the measurement solution is orange-red to red in color.
The method is analogous to APHA 3500-Al B and DIN ISO 10566.
Measuring range
Applicable sample
Soil samples
Reagents, instruments, and materials
 Test/Reagent kit(s)
For the measurement one of the following Spectroquant® test kits is necessary: 
  • Spectroquant® Aluminum Cell Test (1.00594)
  • Spectroquant® Aluminum Test (1.14825)
Instrument(s) and devices
For the measurement one of the following Spectroquant® photometers is necessary:
  • Spectroquant® VIS Spectrophotometer Prove 100 plus (1.73026) 
  • Spectroquant® UV/VIS Spectrophotometer Prove 300 plus (1.73027)
  • Spectroquant® UV/VIS Spectrophotometer Prove 600 plus (1.73028)
  • Spectroquant® Colorimeter Move 100 (1.73632)
Note: Also, legacy Spectroquant® instruments are suitable.
Software for data transfer
Optional Spectroquant® Prove Connect to LIMS software package (Y.11086) to transfer your data into an existing LIMS system.
Instrument accessories
If using Spectroquant® Aluminum Test (1.14825), choose one of the following cells depending on the measuring range:
  • Rectangular cell 10 mm (1.14946) 
  • Rectangular cells 20 mm (1.14947) 
  • Rectangular cells 50 mm (1.14944) 
Other reagents and accessories
  • Hydrochloric acid 25 % for analysis (1.00316)
  • Hydrochloric acid 1 mol/l for analysis (1.09970)
  • Sodium hydroxide solution 10 % for analysis (1.05588)
  • MQuant® Universal Indicator strips pH 0 – 14 (1.09535)
  • MQuant® Indicator strips pH 0 – 6 (1.09531)
  • Water for analysis (1.16754)
  • Charcoal activated for soil tests
  • Analytical balance
  • pH meter
  • Standard laboratory glassware (e.g., glass beakers) and pipettes
  • Spatula
  • Stirring plate or shaker
  • Folded filter
Analytical procedure
Sample preparation
  • In a glass beaker place 10 g of air - dried and sieved (mesh 2 mm) sample with 100 mL hydrochloric acid 1 mol/L and a spatula - tipfull of charcoal activated for soil tests and mix well for 1 hour. The pH - value should be 1 – 2 (check with MQuant® Indicator strips). If necessary, adjust with hydrochloric acid 25 %.
  • Afterwards adjust the pH value of the suspension to 3.0 with sodium hydroxide solution 10 %, using the pH meter.
  • Then remove the undissolved matter by filtration through a folded filter and wash the filter residues 3 times with water for analysis.
  • Collect the filtrate and the washings in a 200-mL volumetric flask, finally fill up to volume with water for analysis and mix well.
Using cat. no. 1.00594: procedure and measurement
For more information on the measurement, see the packaging insert for the test.
Procedure
  • Pipette 6.0 mL pretreated sample into a reaction cell and mix.
  • Add 1 level blue microspoon (in the cap of the Al-1K bottle), close the cell tightly, and shake vigorously until the reagent is completely dissolved.
  • Add 0.25 mL Reagent Al-2K with a pipette and mix.
  • Leave to stand for 5 min (reaction time), then measure the sample in the photometer.
Measurement
  • For photometric measurement the cells must be clean. Wipe, if necessary, with a clean dry cloth.
  • Measurement of turbid solutions yields false-high readings.
  • Aluminum-free samples turn orange on addition of reagents Al-1K and Al-2K.
  • The pH of the measurement solution must be within the range 5.5 - 6.0.
  • The color of the measurement solution remains stable for 15 min after the end of the reaction time stated above.
Hints for measurement
  • It is recommended to zero the method each new working day. To do this, open the method, either by manually selecting the method or by inserting a barcoded cell. Tap the <Settings> button and select the <ZERO ADJUSTMENT> menu item. After prompting, insert the 16 mm zero cell through the corresponding opening. The zero adjustment is performed automatically. Confirm the performance of the zero-adjustment procedure by clicking on <OK>.
  • After the zero has been performed, insert the barcoded Spectroquant® round cell through the corresponding opening, ensuring that the white position mark on the cell is aligned with the positioning mark on the spectrophotometer. The measurement starts automatically.
  • Read off the result in mg/L from the display.
Hint: The above written measurement description is only valid for the Spectroquant® Prove (plus) series photometer. If a different instrument is used, please consult the corresponding instrument manual for more details on how to perform the measurement.
Using cat. no. 1.14825: procedure and measurement
For more information on the measurement, see the packaging insert for the test.
Procedure
  • Pipette 5.0 mL pretreated sample into a test tube.
  • Add 1 level blue microspoon (in the cap of the Al-1 bottle) Reagent Al-1 and shake vigorously until the reagent is completely dissolved.
  • Add 1.2 mL reagent Al-2 with pipette and mix.
  • Add 0.25 mL reagent Al-3 with pipette and mix.
  • Leave to stand for 2 min (reaction time), then fill the sample into the cell, and measure in the photometer.
Note:
  • For measurement in the 50-mm cell both the sample volume as well as the quantities of reagents Al-1, Al-2, and Al-3 must be doubled. Alternatively, the semi-microcell (1.73502) can be used.
  • When using the 50-mm cell, perform the measurement against a separately prepared blank (preparation as per measurement sample, but with distilled water instead of sample). Configure the photometer for blank measurement. 
Measurement
  • Certain photometers may require a blank (preparation as per measurement sample, but with distilled water instead of sample).
  • For photometric measurement the cells must be clean. Wipe, if necessary, with a clean dry cloth.
  • Measurement of turbid solutions yields false-high readings.
  • Aluminum-free samples turn orange on addition of reagents Al-1, Al-2, and Al-3.
  • The pH of the measurement solution must be within the range 5.5 - 6.0.
  • The color of the measurement solution remains stable for 15 min after the end of the reaction time stated above.
Hints for measurement
  • It is recommended to zero the method for each new working day. To do this, open the method by inserting the barcode, tap the <Settings> button and select the <ZERO ADJUSTMENT> menu item. Fill the same cell which will be used for the sample measurement with distilled water. After prompting, insert the filled rectangular cell into the cell compartment. The zero adjustment is performed automatically. Confirm the performance of the zero-adjustment procedure by clicking on <OK>.
  • After the zero adjustment, fill the measurement sample into the same or a matched rectangular cell and insert the cell into the cell compartment. The measurement starts automatically.
  • Read off the result in mg/L from the display.
Hint: The above written measurement description is only valid for the Spectroquant® Prove (plus) series photometer. If a different instrument is used, please consult the corresponding instrument manual for more details on how to perform the measurement.
Analytical quality assurance
Analytical quality assurance (AQA) is recommended before each measurement series.
To check the photometric measurement system (test reagent, measurement device, handling) and the mode of working, the Aluminum standard solution (see section 5 of the respective instruction leaflet) or Spectroquant® CombiCheck 100 can be used. Besides a standard solution with 0.40 mg/l Al, CombiCheck 100 also contains an additional solution for determining sample dependent interferences (matrix effects).
Sample-dependent interferences (matrix effects) can be determined by means of standard addition or dilution.
To view additional notes, visit SigmaAldrich.com/qa-test-kits.
Calculation
Aluminum content in mg/kg Al = analysis value in mg/L Al x 20
Related products
Test kits
Loading
Instruments, software and accessories
Loading
Reagents and test strips
Loading
Reference materials
Loading
References
1.
Lokhanina S, Trubacheva L, Ivanova M, Trubachev A. 2025. Spectrophotometric Determination of Exchangeable (Mobile) Aluminum in Soil Using Arsenazo I. Fine Chemical Engineering.46-52. https://doi.org/10.37256/fce.6120255406