MilliporeSigma
HomeBiopharma Plant Design and Single-Use TechnologiesStreamlining computer system validation (CSV) in bioprocessing
Streamlining computer system validation (CSV) in bioprocessing
To improve product quality and process efficiency, pharmaceutical manufacturers increasingly build interconnected production environments with equipment, process analytical technologies, data analytics and control software, cloud applications, and enterprise systems that must share data reliably and securely. Validating these systems requires a multi-dimensional Computer System Validation (CSV) to meet relevant regulatory requirements.
CSV is the validation of computerized systems to ensure regulatory compliance, correct operation, and data integrity throughout the manufacturing lifecycle. As pharmaceutical facilities become increasingly connected, automated, and digitalized, CSV has grown more complex.
To streamline CSV, organizations need solutions that combine bioprocess expertise, digital accelerators, and process enablers. By adopting emerging methodologies such as computer software assurance (CSA), a risk-based approach to validation, and leveraging vendor expertise, manufacturers can address complexity and maintain confidence in product quality and data integrity.
Read our Bioprocess Systems Services brochure to learn more about our validation support.
Key takeaways
  • Computer system validation (CSV) is a documented process for assuring that computerized systems meet regulatory requirements and consistently and reliably perform as intended.
  • As more manufacturing systems become software-enabled, CSV has emerged as a key element of a commissioning, validation, and qualification (CQV) lifecycle, reflecting the diversity of technology platforms and the complexity of software life cycles in manufacturing environments.
  • Regulatory authorities require computerized systems used in GMP environments be validated to ensure data integrity (that is, that they follow ALCOA+ principles) and security. CSV maps applicable regulatory requirements to specific system functionality and controls (e.g., system access controls using unique user logins and passwords).
  • Best practices for managing CSV complexities include a well-defined user requirements specification (URS), considering the complete chain of custody for critical data, lifecycle management, and implementing risk-based approaches.
  • Historically, CSV relied on extensive documentation and scripted testing for nearly every software feature. In response, the FDA has introduced computer software assurance (CSA), a modern, risk-based framework.
What is CSV in pharma and what are its core requirements?
In regulated pharmaceutical manufacturing environments, CSV is a documented process of assuring that computerized systems consistently meet regulatory and functional requirements and are fit for their intended use.
Key elements of a comprehensive CSV program include:
  • Regulatory compliance: Demonstrate compliance with applicable regulations and industry guidance around the use of computerized systems in GMP environments
  • Risk assessment: Evaluate the potential impact of system failures on product quality, patient safety, and data integrity to determine appropriate validation and control strategies
  • Requirements definitions: Establish clear user requirements, intended use, critical workflows, user roles, and operational expectations that serve as the foundation for validation activities
  • Functional and design specifications: Document how system functionality, configuration, and technical specifications satisfy user and regulatory requirements
  • Data integrity: Verify that data is generated, captured, transferred, stored, and maintained accurately and reliably, with access controls, audit trails, change control processes, and security measures in place
  • Validation documentation: Develop and maintain the records needed to demonstrate compliance including validation plans, requirements specifications, risk assessments, test protocols, test results, traceability matrices, deviations, and final reports
  • Change control: Manage software updates or changes to hardware, configuration changes, integrations, patches, and enhancements through a controlled process that assesses risk and maintains the system's validated state
  • Training and competency: Ensure personnel are properly trained to operate, administer, maintain, and support the system in accordance with established procedures
  • Periodic review and lifecycle management: Conduct regular reviews to verify that systems continue to operate as intended, remain compliant with evolving regulations, and support ongoing business and operational needs
Where does CSV fit within CQV in pharmaceutical manufacturing?
CSV is a component of the broader commissioning, qualification, and validation (CQV) framework used throughout the pharmaceutical industry.
CQV provides documented evidence that facilities, equipment, utilities, and processes are designed, installed, operating, and maintained in a manner that consistently supports product quality and patient safety.
The CQV process begins with commissioning, which verifies that a system is operational and safe prior to formal qualification. Activities such as factory acceptance testing (FAT), site acceptance testing (SAT), startup testing, calibration verification, and utility checks confirm that equipment and systems are correctly installed, functioning per design intent, and ready for formal qualification.
Next, qualification establishes that a system is fit for its intended use. Installation qualification (IQ) verifies that equipment is correctly installed, operational qualification (OQ) confirms it operates within defined parameters, and performance qualification (PQ) demonstrates consistent performance under routine operating conditions.
Finally, validation confirms that processes consistently produce quality products while meeting regulatory requirements. Validation activities may include process validation, cleaning validation, and analytical method validation.
A strong CQV program helps reduce contamination risks, batch failures, regulatory observations, and startup delays. However, achieving and maintaining compliance requires significant time, documentation, and resources, and validation activities can become a critical path item that impacts facility startup, technology deployment, and commercial manufacturing readiness.
Why is CSV a challenge to manage?
As manufacturing systems become software-enabled, CSV has emerged as a core element within the overall CQV lifecycle. In addition to validating system functionality, CSV must also demonstrate regulatory compliance by mapping software features, such as audit trails, access control, or immutable records, to regulations around data integrity, electronic records and signatures, and the use of computerized systems in GMP environments.
Contributing to CSV complexity, pharmaceutical manufacturing facilities operate with equipment, automation platforms, manufacturing execution systems (MES), data historians, LIMS, and enterprise software from multiple suppliers. These systems must exchange information reliably while maintaining compliance with regulations such as 21 CFR Part 11 (electronic records and signatures), 21 CFR Part 210, 211, 212 (data integrity), EudraLex Volume 4 Annex 11 (computerized systems in a GMP environment), and ISPE’s industry guideline Good Automated Manufacturing Practice 5 (GAMP® 5).
Throughout a system’s operational life, software security updates, feature releases, and bug fixes can trigger additional testing, documentation updates, and partial revalidation activities to ensure systems remain in a validated state.
The regulatory burden can be substantial. Validation teams must spend their time creating, reviewing, executing, and maintaining compliance documentation including IQ, OQ, and PQ protocols, traceability matrices, deviations and corrective and preventive actions (CAPAs) records. In many organizations, these activities still rely heavily on manual processes, spreadsheets, and paper-based approvals.
Engineering, quality assurance (QA), manufacturing, automation, information technology, vendors, and validation teams must work together to define requirements, review the system design before executing validation testing, resolve issues, and maintain compliance.
As facilities become more connected and digitalized, cybersecurity concerns add another layer of complexity to validation. Organizations must protect systems from unauthorized access and cyber threats that could impact product quality or patient safety which requires validating controls for secure access, backup and recovery, patch management, and network security.
As a result, CSV requires significant time, resources, and specialized expertise. Experienced validation engineers and QA reviewers are in high demand, creating staffing constraints for many organizations. Despite this, robust CSV programs are essential for ensuring compliance, protecting data integrity, and maintaining confidence in manufacturing operations.
Best practices for managing CSV complexity
While validation requirements grow, organizations can take practical steps to streamline CSV activities without compromising compliance.
Establish clear requirements from the outset. Effective validation begins with a well-defined user requirements specification (URS) that captures not only system functionality, but also intended use, user roles, data integrity and security controls. Clear requirements make it easier to develop traceability, execute testing, and demonstrate compliance.
Understand how data moves across the manufacturing environment. Data may originate in one system, flow through multiple applications, and ultimately support manufacturing decisions elsewhere in the process. Organizations should therefore think beyond individual systems and consider the complete chain of custody for critical data, from origin to final use, to ensure integrity is maintained.
Risk management is central to efficient validation. Historically, organizations adopted a "test everything" approach because of uncertainty about regulatory expectations resulting in excessive documentation, lengthy timelines, and higher validation costs. A risk-based methodology focuses validation efforts on the software functions that have the greatest impact on product quality, patient safety, and data integrity.
Involve equipment and software vendors early is your validation process. While third-party validation consultants possess broad validation expertise, equipment suppliers understand their technologies in detail, including design, configuration, integration, and maintenance. Their involvement accelerates validation, reduces testing effort, and strengthens confidence in critical functionality assessment.
Don’t forget to account for software lifecycle management. Validation doesn’t end at release - software updates, patches, cybersecurity enhancements, configuration changes, and regulatory expectations can affect system performance and compliance. Organizations should establish lifecycle management processes that evaluate changes, assess risk, monitor regulatory updates, and implement appropriate measures to keep systems in a validated state throughout their operational life.
Ensuring data integrity throughout the validation lifecycle
Data integrity is central to CSV requirements. Regulators expect manufacturers to demonstrate that electronic records are trustworthy, complete, and protected from unauthorized modification. This requires understanding how data is generated, transferred, stored, and used throughout the manufacturing process.
Best practices include verifying audit trails which means a secure, computer-generated, time-stamped electronic record, ensuring appropriate user access controls are in place, demonstrating that data is complete, consistent, and accurate, and confirming backup and restore function as intended. Cybersecurity controls should also be evaluated to prevent unauthorized access or data manipulation.
Data integrity is rarely a single-system concern. Information passes between equipment, software platforms, historians, and enterprise systems before being used in operational decisions. Validation programs should assess not just individual systems, but also the integrations connecting them.
Why is risk management important to CSV?
Risk management is the foundation of effective validation because not all software functions carry equal impact. A production system may have hundreds of features, but only a subset directly influences product quality, patient safety, or data integrity. Testing every feature with the same rigor wastes resources without necessarily improving compliance or protecting patients. Critical functions should receive more extensive testing and documentation; lower-risk capabilities only require proportionate effort. This reduces validation burden, accelerates technology deployment and facility startup, and speeds time-to-market.
Risk management also enables continuous improvement. As manufacturers gain operational experience, they can better identify recurring failure modes and higher-risk areas across facilities, continuously refining validation strategies in alignment with ICH Q9 Quality Risk Management principles.
As software environments have become more sophisticated, regulators have formalized these risk management principles into a structured compliance framework, computer software assurance (CSA).  CSA shifts focus from documenting every activity to generating sufficient objective evidence that a system is fit for its intended use. For instance, a single platform such as an MES or LIMS may contain both high and lower-risk functions, each treated proportionately. High process risk functions require scripted testing and thorough documentation; lower-risk functions can be assured through exploratory testing or automated testing. Organizations can also use supplier documentation and objective evidence where appropriate.
The evolution from CSV to CSA and what it means for manufacturers
The CSA framework follows five structured steps: 
  1. Defining intended use
  2. Determining risk
  3. Selecting commensurate assurance activities
  4. Establishing fit-for-purpose records
  5. Managing changes through the same risk-based lens throughout the operational lifecycle.  
This gives pharmaceutical validation teams a clear, auditable methodology.
This shift is particularly important as pharmaceutical manufacturers increasingly adopt cloud-based platforms, SaaS applications, digital batch records, Agile development, DevOps, and continuously updated digital systems — environments where traditional CSV is difficult to sustain. FDA guidance explicitly extends CSA to automation tools, AI/ML tools, and data analytics platforms used in GMP environments, providing a clear regulatory pathway for validating the emerging technologies central to modern pharmaceutical manufacturing.
CSA does not eliminate an organization's compliance responsibilities. What CSA changes is how evidence of that confidence is generated — less burden for lower-risk functions, more focused effort where patient safety and product quality are genuinely at stake. The practical benefits of adopting CSA are compelling: a significant reduction in documentation effort, faster system implementation timelines, lower cost of managing software changes in continuous deployment environments, and stronger inspection readiness.
CSV and lifecycle management
Validation does not end at release and goes beyond software lifecycle management. Manufacturers should also conduct periodic reviews at defined intervals to confirm systems continue to meet their intended use, and that no undocumented changes have occurred. This is distinct from change control.
As personnel join, leave, or change roles, access privileges can drift out of alignment with compliance requirements. Segregation of duties must be enforced, access rights reviewed regularly, and permissions promptly revoked when no longer appropriate. Audit trail review and continuous monitoring are equally essential to investigate anomalies. Under CSA, ongoing performance monitoring can serve as a continuous assurance activity for high-risk functions, helping detect issues before they affect product quality.
When systems are upgraded or replaced, consider data migration as a formal validation activity. Migrated data must be verified for accuracy, completeness, integrity, and traceability, with documented test strategies and post-migration evidence. When systems reach end of life, formal decommissioning requires the documented archiving of GMP records including how secure data is handled, and how transitions are planned to prevent compliance gaps.
Finally, a documented strategy needs to be in place for transitioning legacy CSV-validated systems to CSA principles over time, prioritized by risk. Lifecycle management is a continuous practice — organizations that manage it as an integrated program, rather than isolated tasks, are better positioned for compliance, inspection readiness, and controlled manufacturing operations.
How partnering with experts accelerates CSV
Successful validation requires more than regulatory knowledge. It also requires an understanding of how manufacturing processes, equipment, software, and data systems work together in production environments.
Our CSV experts combine bioprocessing expertise with validation experience to help manufacturers address both the technical and compliance challenges associated with computerized systems.
Unlike general validation consultants, our teams understand the equipment, software, and workflows used in bioprocessing operations. We help customers turn requirements into practical validation strategies, identify critical risks, and streamline testing effort.
Our experts can support customers throughout the validation lifecycle, from requirements, development and risk assessments through qualification, testing, documentation, and ongoing lifecycle management.
Ready to optimize your CSV strategy?
Whether you are implementing a new manufacturing system, expanding a facility, or exploring CSA-based validation, our experts can help. Download our brochure to read more about our validation services or connect with our CSV specialists to discuss your project requirements.
Frequently asked questions
Risk management helps organizations focus validation efforts on software functions that most affect product quality, patient safety, and data integrity. Prioritizing higher-risk functions, reduces unnecessary testing and documentation while maintaining compliance and improving validation efficiency.
The best approach combines risk assessment, root-cause analysis, and change management. By identifying recurring issues and their impact across systems and facilities, organizations can address vulnerabilities and support continuous improvement rather than reacting to failures.
Validation should evaluate data across its lifecycle, from creation and transfer to storage and archival. Best practices include verifying audit trails, user access controls, electronic signatures, backup and recovery processes, cybersecurity protections, and system integrations to ensure data remains complete, consistent, and accurate.
Requirements should clearly define expected functionality, user roles, operating conditions, outcomes, performance expectations, security controls, data handling requirements, and regulatory considerations. Clear requirements reduce ambiguity and make validation activities easier to execute and maintain.
Organizations can streamline documentation with standardized templates, reusable validation content, and risk-based validation. Using supplier documentation and validation evidence can reduce redundant testing, while digital validation tools and centralized document management improve efficiency and consistency.
Continue exploring the investment project journey
Flexible manufacturing process review
Service Supporting
Consider options for designing flexible manufacturing 
No one-size-fits-all solution exists for process and plant layout. Evaluate the different options for future-ready manufacturing. 
Learn more
Ensure compliant and deliverable URS
Service Supporting
Ensure compliant and deliverable URS
Learn how to create compliant, fit-for-purpose URS that addresses the complexity and interdependence of modern manufacturing. 
Learn more
Strengthen single-use supply chain resilience against disruption
Technical Article
Strengthen single-use supply chain resilience against disruption
Learn how we’re expanding our single-use manufacturing network to ensure reliable, high-quality regional production. 
Learn more