MilliporeSigma
HomeReaction Design & OptimizationAdQPhos and Pd pre catalysts for alpha arylation reaction
Substituted Adamantyl Phosphinoferrocenes (AdQPhos) and Precatalysts for Sustainable α-Arylation Reaction in Water
Abstract
Novel polyarylated admantylphosphino ferrocene (AdQPhos) and its Pd-precatalysts were synthesized in multi-gram quantities in excellent yields and purity. This catalytic system demonstrated remarkable activity and selectivity towards palladium-catalyzed α-arylation reactions, accommodating a wide range of nucleophiles, including nitroalkanes, nitriles, amides, esters, and ketones. Compared to the other state-of-the-art catalysts, this system exhibited superior performance in all these aforementioned transformations. Furthermore, by integrating micellar catalysis, these catalysts enabled such carbanion-mediated C-C couplings under mild aqueous conditions, mitigating the need for hazardous solvents like 1,4-dioxane (D201863) or NMP (M79603). Notably, this work also presents the first report for the α-arylation reactions involving cyclic amides under aqueous conditions. Interestingly, the α-arylations of esters proceeded smoothly without employing Zn-enolates (Referomosky reagents), addressing the step-efficiency challenges associated with these reactions. In addition, challenging α-arylations of 3o enolates as nucleophiles to synthesize 4o carbon centers were also demonstrated with the AdQPhos-based catalytic system. A pentagram scale example demonstrated the scalability of this reaction.
Section Overview
Introduction  
Palladium-catalyzed α-arylation reaction has emerged as an important class of reactions primarily due to their ability to forge C-C bonds between heavily functionalized molecules.1–8  Such transformations are extremely relevant in the synthesis of APIs,2,3 natural products,4,5 and agrochemicals.6 Mechanistically, these reactions proceed through the traditional Pd(0)/Pd(II) cycle, where, the transmetalating nucleophile is typically derived from an enolate.9 This approach gives a facile and convenient route to access α-arylated carbonyl compounds, compared to the traditional multi-step cyclization strategies or SNAr reactions carried out under harsher conditions.10,11 The first generation variants of these reactions either employed preformed enolates12,13 or Zn-reagents (Reformatsky reagent)14 as the nucleophiles. This approach was extremely step-inefficient and raised considerable operational difficulties, especially during the scale-up. To overcome some of these challenges, independent work from Buchwald15 and Hartwig16 showcased the utilization of in situ generated enolates from the respective carbonyl compounds. Since then, the field has expanded significantly with the respect to the range of enolate precursors, encompassing aldehydes,17 amides,18 esters,18–20 acids,21 acid chlorides,22 nitroalkanes,23 nitriles,24 and more.9 Process chemists have explored this technology at scale for the synthesis of various APIs and agrochemicals.2,3
Despite significant progress in Pd-catalyzed α-arylation of carbonyl compounds, several key challenges remain (Figure 1). A major limitation is the lack of a general and versatile ligand/catalytic system capable of efficiently accommodating a broad range of nucleophiles. This substantially impacts the overall process efficiency, as chemists must dedicate a significant amount of time and resources into identifying an optimal ligand/catalytic system for each desired transformation. Additionally, α-arylations involving tertiary-carbon centered nucleophiles remain poorly accommodated in this class.23 This is primarily due to the inability of the traditional catalytic systems to facilitate the transmetallation of the sterically challenging nucleophiles or the reductive elimination of coupling partners from the Pd-center.24,25 As a result, these reactions often require elevated temperatures or sophisticated ligands/precatalysts to proceed.26 In addition to substrate and catalyst limitations, scalability remains a major concern, with many reported protocols proving difficult to reproduce under preparative or industrial conditions. Moreover, the prevailing reaction conditions rely heavily on using anhydrous hazardous organic solvents, such as dioxane, NMP, and benzene. The resulting environmental and safety issues, compounded by increasingly stringent global regulations, adversely affect the widespread adoption of this methodology for bulk chemical production.
A representation of a palladium-catalyzed α-arylation reaction of carbonyl compounds. At the top, two reactants are shown: a black and grey sphere representing an aryl halide (with a Br atom) and a blue sphere connected to a CH₂ group bearing an electron-withdrawing group (EWG). The reaction leads to the formation of a new carbon–carbon bond, with the product displaying the blue sphere now bonded to the black sphere, and the EWG retained on the adjacent carbon. Below this reaction scheme, a variety of electron-withdrawing groups are listed in blue text: -NO₂, -CN, -COR, -COOR, -COONR₂, and -CHO. The bottom half of the image displays two structurally complex organic molecules. The left structure includes a TIPS-protected oxygen (labeled in blue), a pyridine ring, a fused bicyclic ring system, and a difluorophenyl ketone, with the newly formed stereogenic center and bond highlighted in pink. The right structure shows a tricyclic framework containing an indole motif, an amide, and a fused ring system, also with the newly formed bond highlighted in pink. The consistent use of pink highlights indicates the key bond formed via α-arylation.
Figure 1.Pd-Catalyzed α-arylation reactions.
Notably, modern sustainable chemistry practices have identified these hazardous solvents as the major contributor to the overall waste generation and pollution. From a pharmaceutical industry perspective, about 85% of the mass employed for an API synthesis is associated with solvent consumption.27 In addition, many of these solvents also pose substantial health risks to workers.28  For instance, NMP (M79603) and DMF (D158550) are reported to be highly toxic to the human reproductive system.28 Due to these reasons, various environmental agencies are bringing stronger regulatory legislations to limit their use unless it is highly necessary. This forces chemists to find greener alternatives.29,30 One prevailing approach in this field is to employ water as the bulk reaction medium by following mother nature’s lead.31,32 Over the last two decades progress has been made in this area by employing water as the bulk reaction medium, primarily enabled by micellar catalysis.33,34 These micelles act as the “nanoreactors” in the bulk medium, facilitating a multitude of synthetic transformations under milder aqueous conditions.33,35 The pioneering contributions by Lipshutz,36,37 Handa,38,39 Gallou,40,41 Kobayashi,42 and others43,44 have substantially advanced the field. Increasing adoption of this technology by agrochemical and pharmaceutical industries reflects its importance in the context of sustainability and green chemistry.45,46
Hence, our goal is to leverage the use of water as a solvent for palladium-catalyzed α-arylations under milder conditions to improve sustainability and safety. Conceptually, employing water as the reaction medium is not ideal for these transformations as the carbanion or related similar intermediates are unstable under aqueous conditions. Recent independent reports from the groups of Handa and Lipshutz demonstrated that the “shielding effect” of micelles can extend the life span of these sensitive intermediates under aqueous conditions and effectively enable the desired transformations.47–50 This was demonstrated in the α-arylations of nitroalkanes,49 phenyl acetonitriles,47 and ketones.50 Careful evaluation of these reports revealed the exhaustive and time consuming screening studies were performed in each of these α-arylations to identify the ideal ligand/catalytic systems. For instance, while the cationic complex t-BuXPhosPd(crotyl)OTf, developed by Colacot et.al.,51 was demonstrated as the most effective catalyst in α-arylations of nitroalkanes, XPhosPd(crotyl)Cl developed by the same group, was used as a “nanocatalyst precursor for α-arylations of nitriles,51 and the highly air sensitive [PdP(t-Bu)3Br]2 was concluded as the most active catalyst in α-arylations of ketones.50 These examples demonstrate the first challenge discussed earlier: “unavailability of a versatile ligand/catalyst system for these reactions.” Therefore, it is an important need to develop a general ligand capable of accommodating a wide range of nucleophiles for all classes of α-arylations under milder conditions, although we are aware that there is not a universal catalyst for every system. However, such a leap could add considerable value to synthetic chemists not only in academia but also for those who want to explore this technology for real-world applications. 
A comprehensive literature evaluation in this direction unveiled the progression of ligands employed in these reactions, ranging from the bidentate BINAP (481084) and XantPhos (526460) to the more advanced Buchwald's XPhos (638064) and RuPhos (663131),52,53 as well as Hartwig's QPhos (675784) ligands.54,55 Among these, the substitution patterns on Hartwig's QPhos are notable, as such a polyarylated phosphino-ferrocene exhibited excellent activity toward a broad range of cross-coupling reactions.
A three-dimensional molecular structure featuring a central transition metal atom coordinated to multiple ligands. The structure includes several phenyl rings and bulky substituents, characteristic of a sterically demanding phosphine ligand. Two large ligands are connected through phosphorus atoms, each bonded to aromatic rings and additional substituents, creating a highly congested coordination environment. The metal center exhibits a square planar or distorted tetrahedral geometry, and it is bound to both phosphorus atoms and to two cyclopentadienyl-like rings that are nearly coplanar. The atomic bonds and elements are represented using ball-and-stick notation, with the metal-ligand bonds clearly highlighted.
Figure 2.Thermal ellipsoid plot of the AdQPhos ligand.
Given our work in developing ferrocene-based ligands, we decided to develop a new catalyst using the polyarylated ferrocene core.56 Modern ligand designs have demonstrated that the incorporation of bulky adamantyl groups on the phosphine has resulted in remarkable enhancement of catalytic properties, as evidenced by Buchwald's findings with AdBrettPhos,57 Beller's studies on CatacXium,58 Stradiatto's research on MorDalphos derivatives,59 Carrow's work on Ad3P60 and our recent report on MPhos.56 By combining adamantyl technology with the uniqueness of the QPhos Cp core, we envisioned that adamantyl phosphino ferrocenes (AdQPhos) might exhibit some unique activities in the area of cross-coupling (Figure 3). Moreover, we sought to establish a robust and scalable synthetic protocol to enable the broader application of this ligand class in preparative and industrially relevant cross-coupling processes.
The structure of the bulky biaryl phosphine ligand AdQPhos and its coordination to various palladium precatalysts. On the far left, AdQPhos is shown in a large circular highlight, featuring a ferrocene core with two cyclopentadienyl rings. One of the rings is substituted with multiple phenyl groups and a diphosphine side chain, including two bulky adamantyl groups. Moving to the right, three different palladium complexes are illustrated, each coordinated with the AdQPhos ligand. In the first complex, labeled AdQPhosPdG3/G4. In the second structure, labeled AdQPhosPd(crotyl)Cl, the palladium is bonded to a crotyl group and a chloride, forming an allylic complex. In the third structure, labeled AdQPhosPdG6, palladium is coordinated with a bromide and a trifluoromethylated phenyl moiety. In all complexes, the AdQPhos ligand and palladium centers are highlighted with pink ovals, and the Pd atoms themselves are colored blue, emphasizing their central catalytic role.
Figure 3.AdQPhos and its precatalysts.
Herein, we describe a very convenient and scalable synthesis of a new class of adamantyl phosphino ferrocene (AdQPhos 936189) and its palladium complexes. These compounds exhibited remarkable catalytic activity in various kinds of challenging α-arylation reactions accommodating a broad range of nucleophiles. Studies were conducted in organic solvents and in green aqueous conditions with relatively low Pd loadings. We also explored these precatalysts for the challenging α-arylations of tertiary centers to test their ability to create quaternary centers. The results of the study are summarized below.
Result and Discussion
Phosphine Ligand synthesis
The ligand synthesis started with the lithiation of readily available starting material, bromoferrocene (725242), followed by quenching with Ad2PCl (737267) (Scheme 1, Part (a)). Interestingly, the ligand (1b) was isolated by simple filtration as the product crashed out from the reaction mixture. The corresponding tBu2P ligand synthesis is extremely tedious due to the isolation problems associated with the instability of the intermediate in the solution phase, byproduct formation, and, as a consequence, the purification process typically requires chromatography.54,55 From a process chemistry perspective, our approach is much more process-economical and convenient. Notably, the traditional installation of Ad2P group requires harsh palladium-catalyzed coupling conditions56 or requires stoichiometric or catalytic amounts of copper salt, especially while synthesizing sterically hindered phosphines, i.e., AdBrettPhos (768154) or MorDalPhos (751618).57,59 Further, adopting a similar procedure used for QPhos synthesis,54,55 the penta-arylation of the bottom cp ring of 1a was achieved cleanly under palladium-catalyzed conditions to form the desired product 1b.
A two-part synthetic scheme outlining the preparation of the bulky ligand AdQPhos and its palladium precatalysts. In part (a), the synthesis of AdQPhos is illustrated in two steps. The starting material, a ferrocene derivative bearing a bromine substituent, reacts with n-butyllithium and a phosphine chloride reagent to form an intermediate compound labeled 1a with a phosphine-adamantyl substituent. This step proceeds with 90% yield, noted in parentheses as 92%. In the second step, this intermediate is reacted in the presence of palladium acetate, sodium tert-butoxide, and an aryl chloride to produce AdQPhos, labeled 1b, in 82% yield (78% isolated yield), where the aryl groups are phenyl substituents. Each reagent and intermediate is identified by product numbers. Part (b) shows the derivatization of AdQPhos into a series of palladium precatalysts. AdQPhos is shown at the top center with arrows pointing to four distinct complexes. The first complex, labeled 2a and 2b, incorporates a sulfonamide biphenyl ligand coordinated to Pd with AdQPhos; 2a has a hydrogen substituent and 2b has a methyl group, with yields of 95% and 92%, respectively. The second structure, labeled 2c, is a Pd complex with a trifluoromethylated aryl bromide ligand, formed in 78% yield. The third structure, 2d, is an allyl-type complex, AdQPhosPd(crotyl)Cl, formed in 90% yield. All structures share a ferrocene backbone with phenyl-substituted rings and are ligated to palladium through the AdQPhos ligand, which contains bulky adamantyl substituents on the phosphorus center.
Scheme 1.Synthesis of (a) AdQPhos and (b) AdQPhos precatalysts.
aConditions: bromoferrrocene (5.0 mmol), n-BuLi (5 mmol), Ad2PCl (5.5 mmol), THF (55 mL), -78 - 23 °C, 19 h; 1a (4.45 mmol), NaO-tBu (44.5 mmol), ArCl (445 mmol), Pd(OAc)2 ( 0.225 mmol), 110 °C.
breaction scale was at 30 g of starting material.
Unlike the QPhos synthesis, which typically requires 10 mol% Pd loading,54,55 the synthesis of our AdQPhos required only half of the Pd loading (5 mol%), although the process was not fully optimized.  We would also like to emphasize that the synthetic protocol of AdQPhos does not rely on tedious work-up or purification techniques such as chromatography in comparison to QPhos. In the case of our AdQPhos, the isolation required only simple filtration followed by washing to get analytically pure material. We were also able to scale up the process to 30 g with similar isolated yield in excellent purity. Subsequently, the Pd complexes such as Buchwald's palladacycles G3 936197 (2a), G4 936200 (2b), oxidative addition complex (G6 2c), and Colacot's crotyl 936219 (2d) were synthesized following the literature procedures in good to excellent yields (Scheme 1, Part (b)).51,61 All the ligands and precatalysts were fully characterized by NMR spectrometry, mass spectroscopy, and other analytical methods. The authenticity of the ligand AdQPhos (1b) was further confirmed with a single-crystal X-ray study (Figure 2). 
Application Studies
With AdQPhos and its precatalysts in hand, we initiated our application studies focusing on assessing their efficacy in various α-arylations encompassing challenging nucleophiles, including enolates derived from amides, esters, ketones, nitriles, and nitroalkanes. To provide a comprehensive analysis, we compared the relative performance of AdQPhos-based precatalysts with other state-of-the-art catalysts for each class of reactions. Remarkably, AdQPhos-based precatalysts showed superior catalytic performances, surpassing the activity of other precatalysts in all classes of α-arylations. This accomplishment met our primary objective of developing a general class of ligand/precatalysts for these cross-coupling reactions. Additionally, we achieved our secondary goal, as these reactions demonstrated operational excellence both in traditional organic solvents and environmentally benign aqueous micellar conditions. The key findings in this regard are discussed in the following sections.
α-arylation of oxoindoles
The α-functionalization of cyclic amides, especially oxoindoles, is notably significant, primarily through the α-arylation method, which facilitates direct synthesis of C-3 arylated oxoindoles.62,63 These structural motifs are widely distributed in various natural products and pharmaceutical targets,62,63 including anti-HIV agents,62 and anti-tumor compounds.63 Despite this significance, such palladium-catalyzed C-3 arylations have not been extensively explored in comparison to other types of α-arylations.64–66  
The first such exclusive study on oxoindoles was reported by Willis and co-workers in 2008.64 Later, Buchwald reported an improved approach with the development of the XPhos based catalytic system.65 Until now, only <10 reports are available on this topic despite the significant interest from medicinal chemists. 64–66 In addition, to the best of our knowledge, there are no existing literature reports on the usage of micellar technology for the palladium-catalyzed α-arylation of alkyl or cyclic amides. Taking this as a technological challenge, we decided to explore our AdQPhos based system for these reactions under milder aqueous conditions.
We used 4-bromo-N,N-dimethylaminobromobenzene (3a, 242950) as the model substrate with N-methyl-2-oxoindole (3b, 466921) as the nucleophile with NaOtBu (703788) as the base in conjunction with PS-750-M (911178) as the surfactant with water as the reaction medium (Figure 4). Our rationale behind utilizing PS-750-M micelles vs. other commercially available surfactants such as TPGS-750-M (763896), Brij-30, lies in their higher inner-core polarity,37 which is crucial for solubilizing polar amide enolates. For comparison studies, we performed the standard reaction using various AdQPhos Pd precatalysts, namely AdQPhos Pd G3 2a, AdQPhos Pd G4 2b, AdQPhos Pd G6 2c, and AdQPhos Pd(crotyl)Cl 2d, as well as several other state-of-the-art precatalysts (see Table 1 for the % yields). Gratefully, all the AdQPhos-based precatalysts (2a, 2b, 2c, and 2d) gave the desired product smoothly with excellent conversions. No double arylation side products were observed. The palladacycles 2a and 2b yielded 80% and 86% of the desired product (3c), respectively (entries 1 and 2). As expected, the oxidative addition complex 2c and crotyl variant 2d exhibited exceptional activity, yielding 99% conversion to the desired product with 92% isolated yield (entries 3 and 4). This could be attributed to the easier generation and consumption of active “L1Pd(0)”. In contrast, the QPhos Pd G3 (903027) or crotyl variants gave inferior conversions to the desired product (31% and 50%, respectively, entries 5 and 6). This clearly demonstrates the superior catalytic activity of the adamantyl (Ad) substituted P over the corresponding t-butyl analog. Other state-of-the-art precatalysts, such as XPhos Pd G3 (763381) gave moderate conversion (59%, entry 7), whereas (dtbpf)PdCl2 (701602) and DavePhos Pd G3 (804959), exhibited poor conversions (traces and <5%) under similar conditions (entries 8 and 9). Interestingly, t-BuXPhos Pd G3 (762229) demonstrated better activity with an 83% yield of the 3c under standard conditions (entry 10). Replacing the PS-750-M with another non-ionic surfactant (2 wt% TPGS-750-M aqueous solution) yielded inferior conversions to 3c (< 39%, entry 12).
A palladium-catalyzed cross-coupling reaction for the synthesis of a substituted indolinone derivative. The starting materials are 4-bromo-N,N-dimethylaniline, labeled 3a, and N-methyl-2-oxoindole, labeled 3b. The aniline derivative 3a has a dimethylamino group on the para position of the benzene ring, while the indole derivative 3b features a five-membered lactam fused to a benzene ring, with a methyl group on the nitrogen. The two substrates undergo a coupling reaction in the presence of 2 mol% of an unspecified palladium catalyst, 2 equivalents of sodium tert-butoxide as base, and a 2 weight percent aqueous solution of PS-750-M. The reaction is conducted at 60 degrees Celsius. The product, labeled 3c, is shown with a newly formed carbon–carbon bond between the aryl ring of 3a and the C-3 position of the indolinone ring system of 3b. The product retains the dimethylamino substituent and the lactam moiety, forming a tri-substituted indolinone structure. Reaction conditions and stoichiometry are noted, and all conversions are stated to be determined by proton NMR.
Figure 4.Synthesis of 3c (3-(4-(Dimethylamino)phenyl)-1-methylindolin-2-one) from 4-bromo-N,N-dimethylaminobromobenzene and N-methyl-2-oxoindole with NaOtBu in conjunction with PS-750-M.
Conditions: 3a (0.55 mmol), 3b (0.5 mmol), [Pd] (2 mol %), NaOtBu (1.0 mmol), 1.0 mL 3 wt % aq. PS-750-M, 60 °C, 20 h (all conversions are based on 1H NMR.
The superiority of PS-750-M could be due to the higher polarity of its micellar inner core. From a sustainability perspective, we performed the same reaction under organic solvent conditions (1,4-dioxane). Only 58% conversion was observed with 2d as the catalyst (entry 11), further highlighting the sustainability factor of our AdQPhos-based catalysts in α-arylation of amides.
Further, the scope of this technology was subsequently explored (Figure 5, 3c-3r). While expanding the scope, we ensured to test the activity of the catalysts 2d under both sustainable (aqueous micellar) as well as the non-sustainable (1,4-dioxane) conditions. Under micellar conditions, the reactions with 5-bromoindole (B68607) containing free N-H gave 67% of the desired C-3 arylated product without any side reactions (3d). Whereas under organic solvent conditions, the yield was almost reduced to half (33%). This was primarily due to the unwanted side reaction at the “N” center. The sterically demanding 2-bromomesitylene (B71608) also reacted smoothly with nucleophile 3b, yielding 80% of the desired product 3e. Similar to the previous example, the reaction under non-sustainable conditions only gave 46% of the desired product. Interestingly, when 3b was reacted with the more sterically challenging substrate bromo-tri-isopropylbenzene (637939) the reaction yield remained excellent (92%, 3g). Furthermore, electron-rich aryl halides such as N,N-dimethylbromobenzene, 4-bromomorpholinyl benzene, 4-bromoanisole, 4-bromo tert-butyl benzene were successfully introduced to the C3 position of the oxoindole (3b) under these conditions with moderate-to-excellent yields (3c-92%; 3f-81%, 3i-86%, 3j-90%). While evaluating the solvent effect on this reaction, it became obvious that micellar technology outperformed the traditional dioxane-based reactions, highlighting the effectiveness of our catalyst system under sustainable aqueous micellar conditions. Notably, this catalyst was inefficient in enabling the desired cross-couplings of challenging quinoline substrates (3k, 3l), presumably due to the known catalyst deactivation by quinoline binding. Similarly, naked oxoindoles are not compatible with the current conditions due to the formation of obvious side products in considerable quantities (3m, 3n).
The scope of a palladium-catalyzed α-arylation reaction between N-methyl-2-oxoindole and a variety of aryl bromide substrates using the AdQPhosPd(crotyl)Cl complex as the catalyst. The reaction is carried out under basic conditions with sodium tert-butoxide at 60 degrees Celsius for 20 hours, using either a polymer-supported aqueous medium (PS-750-M in water) or 1,4-dioxane as the solvent. The general reaction setup is illustrated at the top with the two substrates combining in the presence of 2 mol% catalyst, 2 equivalents of base, and solvent, yielding a variety of substituted indolinone products. Each individual product is displayed below, with yields indicated next to them depending on the solvent used. Blue circles represent yields obtained in the aqueous PS-750-M system, and pink circles represent yields in dioxane. Products include a wide range of functional groups on the aryl ring such as dimethylamino, acetyl, methoxy, trifluoromethyl, tert-butyl, and fused polycyclic structures. High isolated yields are observed for many derivatives in the aqueous system, such as 3h at 95%, 3j at 90%, and 3p at 90%. In contrast, some substrates show significantly reduced yields in dioxane, or produce only trace amounts or mixtures of products, such as 3l, 3m, and 3n.
Figure 5.α-arylation of N-methyl-2-oxoindole (3b)a with different aryl halides using the catalyst AdQPhos Pd(crotyl)Cl (2d) in reaction mediums of PS-750-M in water and 1,4-dioxane.
aConditions: Ar-Br (0.55 mmol), Oxoindole (0.5 mmol), [Pd] (2 mol %), NaOtBu (1.0 mmol), 1.0 mL 3 wt % aq. PS-750-M, 60 °C, 20 h, isolated yield.
Moreover, we explored the scope of generation of quaternary centers through this strategy with double arylation of 3b. Compared with the α-arylation of 2° enolates as discussed above, the reactions utilizing 3° enolates as nucleophiles to synthesize 4° carbon centers through the palladium-catalyzed α-arylation reactions are considerably more challenging due to steric factors associated with the enolates, which limit their transmetalation to the palladium center.9 With slightly increased catalyst and base loading (catalyst, 3 mol%; base, 3 equiv.), the α-arylation reactions went smoothly with decent isolated yields of the desired diarylation products (Figure 5, 3o-3r). For instance, irrespective of the electronic nature of the electrophiles, products 3o, 3p, and 3r were isolated with excellent yields (81%, 90%, and 81%, respectively). This approach also accommodated heterocycles like benzothiophene with moderate yield (3r, 70%). 
Other α-arylations
Following the successful implementation of micellar α-arylations for oxoindoles, our research sought to explore the applicability of AdQPhos-based precatalysts in other α-arylations. In this direction, we employed enolates derived from phenyl acetonitriles, nitroalkanes, esters, and ketones. Our objective was to evaluate the catalytic efficacy of these precatalysts in comparison with the established catalysts. Through these extensive comparative studies, we aimed to ascertain the performance enhancements offered by our precatalysts, thereby underscoring their potential significance in advancing the traditional α-arylation methodologies. 
We began our studies by studying the compatibility of phenyl acetonitrile-derived enolates for the Pd-catalyzed α-arylation reactions under aqueous conditions (Figure 6). In this context, pioneering reports from Hartwig's group had set foundations for designing new catalysts and reaction conditions.24 Following these lines, notable works from the Verkade67 and Crudden25 groups improved procedures to accommodate broader substrate scope under milder conditions. Recently, Gessner’s YPhos has shown similar reactivity for the Hiyama-Denmark coupling.68 However, most of these protocols encountered drawbacks:
  • High catalyst loading due to catalyst poisoning due to the Pd-CN binding,
  • Elevated reaction temperature (to promote reductive elimination),
  • Need of stronger bases like LiHMDS (324620) and NaHMDS (235083), and
  • Use of hazardous organic solvents as the reaction medium.
In addressing these challenges, recently, Handa and co-workers reported a micellar nanocatalytic approach based on PS-750-M,47 although the challenges of catalyst recyclability and synthetic reproducibility using nanocatalysis still persist. It is important to note that, based on our literature search, until today, there is not a homogenous catalyst system that could address all of the four challenges mentioned earlier. We were curious to see if our AdQPhos-based system could address some of these challenges.
Three different palladium-catalyzed α-arylation reactions involving aryl bromides and carbon-based nucleophiles under various conditions. In the first reaction, labeled a, 3-methoxyphenyl bromide reacts with 3-methoxyphenylacetonitrile in the presence of 2 mol% catalyst, 2 equivalents of potassium hydroxide, and an aqueous PS-750-M medium at 60 degrees Celsius, yielding a substituted benzonitrile product. In the second reaction, labeled b, 3-methoxyphenyl bromide reacts with 1-nitropropane using 3 mol% catalyst and 1.5 equivalents of dipotassium phosphate monohydrate in dioxane at 60 degrees Celsius, yielding a nitroalkylated arene product. In the third reaction, labeled c, 4-bromo-N,N-dimethylaniline undergoes α-arylation with tert-butyl acetate in the presence of 2 mol% catalyst and 2 equivalents of lithium hexamethyldisilazide (LiHMDS) in toluene at room temperature, yielding a tert-butyl aryl ketone. Each transformation demonstrates a different nucleophile and optimized set of conditions, highlighting the versatility of the catalytic system in forming carbon–carbon bonds with aryl halides across various functionalized substrates.
Figure 6.(a) α-arylation of 3-methoxy phenyl acetonitrile 209392 (4b); (b) α-arylation nitropropane (5a); (c) α-arylation of tert-butyl acetate (6a).
We started our optimization studies employing 3-methoxy phenyl acetonitrile (209392, 4b) as the nucleophile, 4-bromoanisole (B56501, 4a) as the electrophile, KOH as the base, and aqueous micelles of PS-750-M as the reaction medium (Figure 6, Part (a)). To showcase the effectiveness of our AdQPhos-based system over others, we also performed an extensive control study with other catalysts. To our delight, the desired product, 2-(3-methoxyphenyl)-2-(4-methoxyphenyl)acetonitrile 4c, was obtained in excellent yields with palladacycles 2a and 2b (89% and 94%, respectively; Table 2, entries 1 and 2). The reaction was further improved to 98-99% with the crotyl or G6 variants (entries 3 and 4). Inconsistent with the previous optimization studies, the reaction yields dropped substantially while using QPhos palladacycle or its crotyl variants (60% and 82%, respectively; entries 5 and 6). The conversion to the desired product 4c dropped to 59% when the bisphosphine-based (dtbpf)PdCl₂ was used as the catalyst (entry 7). The reaction yields remarkably improved when XPhos Pd G3 was used (88%, entry 8). This comparative study indicated the superiority of AdQPhos-based precatalysts over traditional biaryl or other types of state-of-the-art phosphines. 
Further, the suitability of our AdQPhos systems in the α-arylations of nitroalkane was studied and compared to the traditionally employed catalytic systems (Figure 6, Part (b)).23,69 This particular type of α-arylation has been relatively underexplored, although the resulting products are the key precursors for accessing functionalized secondary/tertiary amines. Previously, the Buchwald and69  Kozlowski23 groups independently reported conditions for this transformation with some critical insights on mechanistic pathways. However, these protocols suffered serious limitations such as (a) higher [Pd] loadings (up to 10 mol%), (b) toxic organic solvent usage, and (c) operational difficulty where the experiments were performed under the glove-box conditions. Later, in 2017, Handa and co-workers reported an improved technology by introducing micellar catalysis.49 However, their method only explored the nitroethane and nitropropanes as the nucleophiles, and the reaction yields with nitropropane were moderate for most substrates studied therein.
In our current study, we started our studies using 4-bromoanisole (4a) and nitropropane (8.06851, 5a) as the model coupling partners with K3PO4 as the base under organic solvent conditions (1,4-dioxane) with lowered catalyst loading (3 mol %) in comparison to those reported (up to 10 mol %). The catalytic activity of the AdQPhos Pd-precatalysts was compared with other state-of-the-art precatalysts. Notably, the desired reaction proceeded smoothly without double arylations when employing AdQPhos precatalysts (2a, 2b, 2c, and 2d) (Table 3). Although under the standard reaction conditions with palladacycles 2a and 2b moderate yields (57% and 59%, respectively) of the desired product 2-(4-methoxyphenyl)butanenitrile 5b (entries 1 and 2) were observed, the G6 or crotyl variants of AdQPhos exhibited excellent activity with an impressive 99% conversion to the desired product (96% isolated yield, entries 3 and 4). Similar to the previous α-arylation with amides, the QPhos-based Pd G3 or crotyl variants only gave <10% of 5b, showcasing the remarkable superiority of our AdQPhos-based catalyst system (entries 5 and 6). Buchwald's XPhos Pd G3 gave moderate conversion (75%), whereas DavePhos Pd G3 gave only traces of 5b (entries 7 and 8). The bidentate ferrocene-based precatalyst, (dtbpf)PdCl₂, also gave poor conversions (28%) under similar conditions (entry 9). Interestingly, t-BuXPhos Pd G3 showed excellent reactivity, resulting in a 97% yield of the desired product 5b under these conditions (entry 10). With the effort to replace the hazardous solvent, 1,4-dioxane in this case, with water using micellar catalysis, we conducted extensive screening studies to identify suitable micellar media. With the micelles of PS-750-M, utilizing 2d as the catalyst, we achieved a remarkably high yield (98%) of 5b under standard conditions (entry 11). In contrast, the use of t-BuXPhos Pd G3 under micellar conditions was ineffective, as the reaction yield dropped to <31% (entry 12) in comparison to the reaction in organic solvent (97%, entry 10). These extensive screening studies demonstrated the superior adaptability and activity of AdQPhos-derived precatalysts over other state-of-the-art catalysts for this class of reaction. 
In order to broaden the scope of nucleophiles in the α-arylations enabled by AdQPhos and its pre-catalysts, we explored esters in this study (Figure 6, Part (c)). Such α-arylations reactions are one of the most challenging classes of reactions due to:
  • Instability of enolate (decomposition to ketene or possible decarboxylation)70,71
  • Ester hydrolysis under the reaction conditions19 
  • Catalyst poisoning due to the possible difficulties in the reductive elimination18
To overcome these challenges, current strategies often employ masked ester nucleophiles such as Reformatsky reagents,18 or, silyl ketene acetals, etc.72,13 These approaches, however, suffered from several limitations, including step inefficiency, reliance on hazardous reagents, and operationally challenging protocols.13,72 Consequently, a catalyst-enabled, economically feasible protocol for α-arylation reactions involving esters remains elusive despite the extensive presence of α-arylated esters in APIs,73,74 and natural products.4,75  In this direction, we started our studies by employing the precatalysts of AdQPhos for the α-arylation of a nucleophile,  tert-butyl acetate (8.02189, 6a) with 4-bromo-N,N-dimethylaminobromobenzene (3a) using toluene as the reaction medium. To evaluate the performance of AdQPhos precatalysts, we compared them with other state-of-the-art catalysts (Table 4). The AdQPhos-derived precatalysts (2a, 2b, 2c, and 2d) gave 39%, 54%, 99%, and 80%, respectively, of the desired product tert-butyl 2-(4-(dimethylamino)phenyl)acetate 6b, without any detectable amounts of possible diarylated product (entries 1-4). It is evident that the Pd G6 version of the AdQPhos ligands showed excellent activity. In contrast, corresponding precatalysts derived from QPhos only gave trace yields (8% and 32 %, entries 5, 6) for 6b. Such a lowered conversion is somewhat anticipated: typically, to enable such reactions using QPhos-based catalysts, the Reformatsky-type reagents are used as the nucleophile. Other ferrocene-based bidentate ligands like dtbpf did not yield any detectable product (entry 7), reiterating the demand for a monodentate phosphine containing an Ad moiety on the phosphorus. We also benchmarked the reaction with other oxidative addition complexes of monophosphines, such as Buchwald's RuPhos Pd G6, which gave only 74% of the desired product (entry 8). 
These extensive comparative studies demonstrated the versatility of AdQPhos-derived precatalysts in enabling α-arylations of a diverse array of nucleophiles as well as their superiority over traditionally employed state-of-the-art Pd-precatalysts. This key observation positions the AdQPhos systems as potentially universal catalysts for Pd-catalyzed α-arylation reactions. Following these studies, the scope of these desired transformations was evaluated under our optimized conditions. We started our substrate scope evaluations with the α-arylations of phenyl acetonitriles (Figure 7, Part (a), 4d-4h). During these studies, we performed the reactions under both aqueous and organic solvent conditions to gain insights into the solvent's influence on each α-arylation reaction. All the reactions proceeded with moderate-to-excellent yields, no dehalogenations or other side reactions were observed with the nitriles containing chloro groups (4d, 82%; 4h, 86%). Heterocycles such as 4-morpholinyl-containing aryl bromides readily reacted with nitriles to yield the desired products in good yields (4d, 82%; 4g, 81%). Electron-rich (4d, 4g) or deficient (4e, 4h) or neutral substrates (4f) participated in the desired reactions without causing any significant drop in yield. It is important to note that these reactions proceeded effectively only under aqueous conditions. Under the conventional organic solvent conditions (e.g. 1,4-dioxane), considerably lower amounts of products were formed (4d - 4h). This observation emphasizes the superiority of our AdQPhos in enabling sustainable Pd-catalyzed cross-coupling reactions by leveraging micellar catalysis.33  
Further, we extended our substrate scope evaluations for the nitro alkane α-arylations. Similar to the nitrile α-arylation, we performed reactions under both aqueous and organic solvent conditions. Conspicuously, the reaction yields were superior under organic solvent compared to surfactant conditions for most substrates, although moderate to good yields were typically obtained from the reactions employing micellar technology. (Figure 7, Part (b), 5b-5f). Nevertheless, our AdQPhos-based catalysts exhibited remarkable versatility, accommodating a wide range of aryl bromides. The electron-withdrawing (5f), and electron-donating (5b-5e) functional group-containing substrates participated in the reaction effectively to yield the desired product in moderate to excellent yield. The reaction exhibited favorable outcomes when dealing with substrates bearing heterocycles (5e).
Four panels of palladium-catalyzed α-arylation reactions involving different classes of nucleophiles—nitriles, nitroalkanes, esters, and ketones—paired with various substituted aryl bromides under different catalytic and solvent conditions. In section a, aryl bromides undergo α-arylation with benzylic nitriles in the presence of AdQPhosPd(crotyl)Cl, potassium hydroxide, and either an aqueous PS-750-M medium or 1,4-dioxane at 60 degrees Celsius. Eight examples are shown with different yields, highlighting a strong solvent effect where PS-750-M consistently outperforms dioxane. Section b shows the α-arylation of nitroalkanes under similar conditions, where three mol% of the same catalyst and dipotassium phosphate are used, again comparing the two solvents. The results vary depending on the substitution pattern on the aryl ring, with generally higher yields observed in aqueous PS-750-M. Section c features the α-arylation of ester enolates using AdQPhosPdG6 and lithium hexamethyldisilazide in toluene at room temperature. The products include arylated esters with dimethylamino, morpholino, and thienyl substituents, with moderate to high yields. Section d illustrates the α-arylation of ketones using AdQPhosPdG4 and either potassium hydroxide or sodium tert-butoxide under aqueous savie or dioxane conditions. Ten product examples are shown, including various aromatic and heterocyclic motifs, with clear solvent-dependent differences in reaction efficiency. The image emphasizes substrate scope and solvent impact across diverse α-arylation pathways using tailored phosphine–palladium catalysts.
Figure 7.(a) α-arylation of nitriles, conditions: Ar-Br (0.6 mmol, 1.2 equiv.), ArCH2CN (0.5 mmol, 1.0 equiv.), [Pd] (3 mol%), KOH (1.0 mmol, 2.0 equiv.), 1.0 mL 3 wt% aq. PS-750-M or 1,4-dioxane 60 °C, 20 h (isolated yield) (b) α-arylation of nitroalkanes, conditions: Ar-Br (0.5 mmol), RCH2NO2 (2.5 mmol), [Pd] (3 mol%), K3PO4 (0.75 mmol), 1.0 mL 3 wt% aq. PS-750-M or 1,4-dioxane 60 °C, 20 h, (isolated yield) (c) α-arylation of esters, conditions: Ar-Br (0.5 mmol, 1.0 equiv.), RCH2COOR (1.0 mmol, 2.0 equiv.), [Pd] (2 mol%), LiHMDS (1.0 mmol, 2.0 equiv.), 2.0 mL toluene, 20 h (isolated yield) (d) α-arylation of ketones, conditions: Ar-Br (0.5 mmol or 1.1 mmole), RCH2COR (0.5 mmol), [Pd] (1 mol%), KOH or NaOtBu (0.75 mmol or 1.1 mmol), 1.0 mL 3 wt% aq. PS-750-M or 1,4-dioxane 60 °C, 20 h (isolated yield).
Next, the scope of α-arylation of esters was briefly evaluated by exploring different combinations of aryl halides and esters (Figure 7, Part (c), 6b-6e). It is important to note that under our reaction conditions, even with excess electrophiles, the reaction exclusively produced monoarylated ester products. Furthermore, these reaction conditions proved to be compatible with ethyl esters (6c, 6e), which are prone to base-mediated hydrolysis under typical reaction conditions. The reaction yields achieved with these conditions ranged from moderate to excellent, highlighting the versatility and effectiveness of our protocol, which accommodated electron-rich and heterocyclic aryl halides as electrophiles effectively. On the nucleophile side, the protocol demonstrated compatibility with both alkyl and benzyl esters. Notably, quinoline-based aryl halides seem to have issues under current conditions. 
Additionally, the catalytic activity of AdQPhos or its precatalysts was evaluated for the α-arylation of ketones (Figure 7, Part (d)). Despite the numerous reports on the utilization of organic solvents, primarily employing hazardous solvents (1,4-dioxane or NMP),76 only a few studies have focused on enabling these reactions under milder and environmentally benign conditions, i.e., using water as the solvent.50 In this regard, recently, Lipshutz and coworkers developed a new technology under aqueous conditions by using [PdP(t-Bu)3Br]2 as a catalyst (up to 1 mol%), aqueous micelles of TPGS-750-M as a reaction medium, and KOt-Bu as the base.50 It is worthwhile to highlight that the Pd(I) dimer of t-Bu3P is one of the most unstable Pd catalysts; hence, it often causes issues in reproducing the results. With our bench-stable AdQPhos Pd G4 (up to 1 mol %), the reactions were carried out in water using a readily available base, KOH in conjunction with a new generation biodegradable surfactant, "Savie" developed by Lipshutz.77 A short substrate scope evaluation was performed to showcase the generality of this versatile catalyst (Figure 7, Part (d)). The reaction proceeded smoothly for cyclic (7a-7c, 7g-7j) and acyclic ketones (7d, 7e) with moderate-to-excellent yields. We continued assessing the solvent effect on catalysis by performing each reaction under aqueous and traditional organic solvent conditions. The coupling between cyclic ketone α-tetralone and sterically challenging 2-bromomesitylene proceeded smoothly to yield the desired product 7a with excellent yield under aqueous conditions (87%). Under organic solvent conditions, the reaction yield was only 71%, with the remaining unreacted starting materials (ketone and aryl halide). Furthermore, this approach was compatible with various heterocycles, including indoles bearing free N-H (7b), benzothiophenes (7c), and benzofurans (7e), with excellent isolated yields. Notably, unlike the α-arylation of nitroalkanes, the reactions on ketones proceeded more efficiently under micellar conditions than under traditional organic solvent conditions for most of the substrates, which, again, demonstrates the sustainability contributions from our AdQPhos-based system in the α-arylation chemistry.
Combining all these studies, we demonstrated that the AdQPhos-based system is a general, practical catalytic system for the palladium-catalyzed α-arylation reactions under organic solvent as well as under green aqueous conditions. 
Scalability and Sustainability Score
To demonstrate the scalability of our AdQPhos-based technology for potential commercial applications, we picked the amide α-arylation reactions and performed them at a pentagram scale.
A gram-scale reaction scheme in which N-methyl-2-oxoindole, labeled as compound 3b and used in a quantity of 5 grams (33.9 mmol), is coupled with 2-bromomesitylene, labeled as compound 3t, under palladium-catalyzed α-arylation conditions. The reaction employs 2 mol% of a catalyst identified as 2d, along with sodium tert-butoxide as the base, in aqueous PS-750-M as the reaction medium, at 60 degrees Celsius and 0.5 molar concentration. The product formed is 3c, a mesityl-substituted oxindole, isolated in 91% yield corresponding to 8.1 grams. A DOZN score of 54 is prominently displayed, indicating an assessment of the reaction's green chemistry profile. The structural formulas for all three compounds are shown, with the core oxindole scaffold and the mesityl aryl group highlighted.
Figure 8.Gram-scale example. Synthesis of 3-mesityl-1-methylindolin-2-one 3e from N-methyl-2-oxoindole 466921 and 2-bromomesitylene B71608.
Conditions: 3b (33.9 mmol), 2-bromomesitlyene (35.7 mmol), AdQPhos Pd(crotyl)Cl (0.678 mmol), NaOtBu (67.8 mmol), 68 mL 3 wt% aq. PS-750-M, 60 °C, 20 h.
The expected α-aylated amide product, 3-mesityl-1-methylindolin-2-one 3e, was isolated in 91% yield, highlighting the usability of our AdQPhos-based system in reaction scale-up. The sustainability score of this reaction was evaluated using DOZN™.78 As the reaction was conducted under micellar conditions, the environmental impact of this transformation was remarkably lower compared to the traditional methods, and this was clearly reflected in the lower DOZN™ score of 54.
Conclusion
In summary, we have developed a new class of polyarylated diadamentylphosphino ferrocene (AdQphos) and its precatalysts as versatile catalytic systems for the Pd-catalyzed α-arylation reactions. The ligand synthesis is simple, scalable, and does not involve tedious workups or purification techniques. The scope and superiority of this catalytic system were demonstrated for a wide variety of nucleophiles, including nitroalkanes, nitriles, amides, and ketones. Most importantly, by incorporating catalysis in water under micellar conditions, the AdQphos-based catalytic system demonstrated the feasibility of enabling α-arylation reactions under milder aqueous conditions by replacing toxic/hazardous solvents such as 1,4-dioxane or NMP with some exceptions, bringing modern organic synthesis closer to being ideally sustainable.
Related Products
Loading
References
1.
Gildner PG, Colacot TJ. 2015. Reactions of the 21st Century: Two Decades of Innovative Catalyst Design for Palladium-Catalyzed Cross-Couplings. Organometallics. 34(23):5497-5508. https://doi.org/10.1021/acs.organomet.5b00567
2.
Desai LV, Hay MB, Leahy DK, Wei C, Fanfair D, Rosner T, Hsiao Y. 2013. Development of a scalable palladium-catalyzed α-arylation process for the synthesis of a CGRP antagonist. Tetrahedron. 69(27-28):5677-5684. https://doi.org/10.1016/j.tet.2013.04.057
3.
Ulysse LG, Yang Q, McLaws MD, Keefe DK, Guzzo PR, Haney BP. 2010. Process Development and Pilot-Scale Synthesis of New Cyclization Conditions of Substituted Phenylacetamides to Tetrahydroisoquinoline-2-ones Using Eaton’s Reagent. Org. Process Res. Dev. 14(1):225-228. https://doi.org/10.1021/op9002533
4.
Sivanandan ST, Shaji A, Ibnusaud I, Seechurn CCCJ, Colacot TJ. 2015. Palladium‐Catalyzed α‐Arylation Reactions in Total Synthesis. Eur J Org Chem. 2015(1):38-49. https://doi.org/10.1002/ejoc.201403301
5.
Nicolaou KC, Bulger PG, Sarlah D. 2005. Palladium-Catalyzed Cross-Coupling Reactions in Total Synthesis. Angew. Chem. Int. Ed. 44(29):4442-4489. https://doi.org/10.1002/anie.200500368
6.
Devendar P, Qu R, Kang W, He B, Yang G. 2018. Palladium-Catalyzed Cross-Coupling Reactions: A Powerful Tool for the Synthesis of Agrochemicals. J. Agric. Food Chem. 66(34):8914-8934. https://doi.org/10.1021/acs.jafc.8b03792
7.
Johansson C, Colacot T. 2010. Metal‐Catalyzed α‐Arylation of Carbonyl and Related Molecules: Novel Trends in CC Bond Formation by CH Bond Functionalization. Angew Chem Int Ed. 49(4):676-707. https://doi.org/10.1002/anie.200903424
8.
Firsan SJ, Sivakumar V, Colacot TJ. 2022. Emerging Trends in Cross-Coupling: Twelve-Electron-Based L1Pd(0) Catalysts, Their Mechanism of Action, and Selected Applications. Chem. Rev. 122(23):16983-17027. https://doi.org/10.1021/acs.chemrev.2c00204
9.
Culkin DA, Hartwig JF. 2003. Palladium-Catalyzed α-Arylation of Carbonyl Compounds and Nitriles. Acc. Chem. Res. 36(4):234-245. https://doi.org/10.1021/ar0201106
10.
Kondoh A, Aoki T, Terada M. 2018. Organocatalytic Arylation of α‐Ketoesters Based on Umpolung Strategy: Phosphazene‐Catalyzed SNAr Reaction Utilizing [1,2]‐Phospha‐Brook Rearrangement. Chemistry A European J. 24(50):13110-13113. https://doi.org/10.1002/chem.201803218
11.
Caubere P, Lalloz L. 1975. Arynic and SNAr reactions of polyhalogenobenzenes. IV. Condensation of ketone enolates. J. Org. Chem. 40(20):2853-2858. https://doi.org/10.1021/jo00908a001
12.
Kosugi M, Suzuki M, Hagiwara I, Goto K, Saitoh K, Migita T. 1982. A NEW PALLADIUM CATALYZED AROMATIC ACETONYLATION BY ACETONYLTRIBUTYLTIN. Chem. Lett. 11(6):939-940. https://doi.org/10.1246/cl.1982.939
13.
Carfagna C, Musco A, Sallese G, Santi R, Fiorani T. 1991. Palladium-catalyzed coupling reactions of aryl triflates or halides with ketene trimethylsilyl acetals. A new route to alkyl 2-arylalkanoates. J. Org. Chem. 56(1):261-263. https://doi.org/10.1021/jo00001a049
14.
Orsini F, Pelizzoni F, Vallarino L. 1989. Influence of the catalyst on the Pd0-mediated reactions of BrZnCH(CH3) COOtBu with vinyl and aryl triflates. Journal of Organometallic Chemistry. 367(3):375-382. https://doi.org/10.1016/0022-328x(89)87059-7
15.
Palucki M, Buchwald SL. 1997. Palladium-Catalyzed α-Arylation of Ketones. J. Am. Chem. Soc. 119(45):11108-11109. https://doi.org/10.1021/ja972593s
16.
Hamann BC, Hartwig JF. 1997. Palladium-Catalyzed Direct α-Arylation of Ketones. Rate Acceleration by Sterically Hindered Chelating Ligands and Reductive Elimination from a Transition Metal Enolate Complex. J. Am. Chem. Soc. 119(50):12382-12383. https://doi.org/10.1021/ja9727880
17.
Martín R, Buchwald SL. 2008. An Improved Protocol for the Pd-Catalyzed α-Arylation of Aldehydes with Aryl Halides. Org. Lett. 10(20):4561-4564. https://doi.org/10.1021/ol8017775
18.
Hama T, Liu X, Culkin DA, Hartwig JF. 2003. Palladium-Catalyzed α-Arylation of Esters and Amides under More Neutral Conditions. J. Am. Chem. Soc. 125(37):11176-11177. https://doi.org/10.1021/ja036792p
19.
Moradi WA, Buchwald SL. 2001. Palladium-Catalyzed α-Arylation of Esters. J. Am. Chem. Soc. 123(33):7996–8002. https://doi.org/10.1021/ja010797
20.
Shaughnessy KH, Hamann BC, Hartwig JF. 1998. Palladium-Catalyzed Inter- and Intramolecular α-Arylation of Amides. Application of Intramolecular Amide Arylation to the Synthesis of Oxindoles. J. Org. Chem. 63(19):6546-6553. https://doi.org/10.1021/jo980611y
21.
Liu X, Hartwig JF. 2003. Palladium-Catalyzed α-Arylation of Azlactones to Form Quaternary Amino Acid Derivatives. Org. Lett. 5(11):1915-1918. https://doi.org/10.1021/ol034570q
22.
Sha S, Zhang J, Walsh PJ. 2015. Palladium-Catalyzed α-Arylation of Aryl Acetic Acid Derivatives via Dienolate Intermediates with Aryl Chlorides and Bromides. Org. Lett. 17(3):410-413. https://doi.org/10.1021/ol503545j
23.
Walvoord RR, Berritt S, Kozlowski MC. 2012. Palladium-Catalyzed Nitromethylation of Aryl Halides: An Orthogonal Formylation Equivalent. Org. Lett. 14(16):4086-4089. https://doi.org/10.1021/ol301713j
24.
Culkin DA, Hartwig JF. 2002. Synthesis, Characterization, and Reactivity of Arylpalladium Cyanoalkyl Complexes:  Selection of Catalysts for the α-Arylation of Nitriles. J. Am. Chem. Soc. 124(32):9330-9331. https://doi.org/10.1021/ja026584h
25.
Nambo M, Yar M, Smith JD, Crudden CM. 2015. The Concise Synthesis of Unsymmetric Triarylacetonitriles via Pd-Catalyzed Sequential Arylation: A New Synthetic Approach to Tri- and Tetraarylmethanes. Org. Lett. 17(1):50-53. https://doi.org/10.1021/ol503213z
26.
Lundgren RJ, Stradiotto M. 2012. Addressing Challenges in Palladium‐Catalyzed Cross‐Coupling Reactions Through Ligand Design. Chemistry A European J. 18(32):9758-9769. https://doi.org/10.1002/chem.201201195
27.
Jimenez-Gonzalez C, Ponder CS, Broxterman QB, Manley JB. 2011. Using the Right Green Yardstick: Why Process Mass Intensity Is Used in the Pharmaceutical Industry To Drive More Sustainable Processes. Org. Process Res. Dev. 15(4):912-917. https://doi.org/10.1021/op200097d
28.
Jordan A, Hall CGJ, Thorp LR, Sneddon HF. 2022. Replacement of Less-Preferred Dipolar Aprotic and Ethereal Solvents in Synthetic Organic Chemistry with More Sustainable Alternatives. Chem. Rev. 122(6):6749-6794. https://doi.org/10.1021/acs.chemrev.1c00672
29.
Clarke CJ, Tu W, Levers O, Bröhl A, Hallett JP. 2018. Green and Sustainable Solvents in Chemical Processes. Chem. Rev. 118(2):747-800. https://doi.org/10.1021/acs.chemrev.7b00571
30.
Sheldon RA. 2019. The greening of solvents: Towards sustainable organic synthesis. Current Opinion in Green and Sustainable Chemistry. 1813-19. https://doi.org/10.1016/j.cogsc.2018.11.006
31.
Chanda A, Fokin VV. 2009. Organic Synthesis “On Water”. Chem. Rev. 109(2):725-748. https://doi.org/10.1021/cr800448q
32.
Butler RN, Coyne AG. 2010. Water: Nature’s Reaction Enforcer—Comparative Effects for Organic Synthesis “In-Water” and “On-Water”. Chem. Rev. 110(10):6302-6337. https://doi.org/10.1021/cr100162c
33.
Ansari TN, Gallou F, Handa S. 2023. Palladium-catalyzed micellar cross-couplings: An outlook. Coordination Chemistry Reviews. 488215158. https://doi.org/10.1016/j.ccr.2023.215158
34.
Lipshutz BH, Ghorai S, Cortes‐Clerget M. 2018. The Hydrophobic Effect Applied to Organic Synthesis: Recent Synthetic Chemistry “in Water”. Chemistry A European J. 24(26):6672-6695. https://doi.org/10.1002/chem.201705499
35.
Cortes-Clerget M, Yu J, Kincaid JRA, Walde P, Gallou F, Lipshutz BH. 2021. Water as the reaction medium in organic chemistry: from our worst enemy to our best friend. Chem. Sci. 12(12):4237-4266. https://doi.org/10.1039/d0sc06000c
36.
Duplais C, Krasovskiy A, Lipshutz BH. 2011. Organozinc Chemistry Enabled by Micellar Catalysis. Palladium-Catalyzed Cross-Couplings between Alkyl and Aryl Bromides in Water at Room Temperature. Organometallics. 30(22):6090-6097. https://doi.org/10.1021/om200846h
37.
Handa S, Andersson MP, Gallou F, Reilly J, Lipshutz BH. 2016. HandaPhos: A General Ligand Enabling Sustainable ppm Levels of Palladium‐Catalyzed Cross‐Couplings in Water at Room Temperature. Angew Chem Int Ed. 55(16):4914-4918. https://doi.org/10.1002/anie.201510570
38.
Kaur G, Kaur K, Handa S. 2022. Efficient catalysis in dynamic nanomicelles of PS-750-M suspended in water. Current Opinion in Green and Sustainable Chemistry. 38100690. https://doi.org/10.1016/j.cogsc.2022.100690
39.
Ansari TN, Taussat A, Clark AH, Nachtegaal M, Plummer S, Gallou F, Handa S. 2019. Insights on Bimetallic Micellar Nanocatalysis for Buchwald–Hartwig Aminations. ACS Catal. 9(11):10389-10397. https://doi.org/10.1021/acscatal.9b02622
40.
Gallou F, Isley NA, Ganic A, Onken U, Parmentier M. 2016. Surfactant technology applied toward an active pharmaceutical ingredient: more than a simple green chemistry advance. Green Chem. 18(1):14-19. https://doi.org/10.1039/c5gc02371h
41.
Lippincott DJ, Landstrom E, Cortes-Clerget M, Lipshutz BH, Buescher K, Schreiber R, Durano C, Parmentier M, Ye N, Wu B, et al. 2020. Surfactant Technology: With New Rules, Designing New Sequences Is Required!. Org. Process Res. Dev. 24(5):841-849. https://doi.org/10.1021/acs.oprd.9b00454
42.
Kobayashi S, Xu P, Endo T, Ueno M, Kitanosono T. 2012. Chiral Copper(II)‐Catalyzed Enantioselective Boron Conjugate Additions to α,β‐Unsaturated Carbonyl Compounds in Water. Angew Chem Int Ed. 51(51):12763-12766. https://doi.org/10.1002/anie.201207343
43.
Mattiello S, Rooney M, Sanzone A, Brazzo P, Sassi M, Beverina L. 2017. Suzuki–Miyaura Micellar Cross-Coupling in Water, at Room Temperature, and under Aerobic Atmosphere. Org. Lett. 19(3):654-657. https://doi.org/10.1021/acs.orglett.6b03817
44.
Kaplaneris N, Vilches-Herrera M, Wu J, Ackermann L. 2022. Sustainable Ruthenium(II)-Catalyzed C–H Activations in and on H2O. ACS Sustainable Chem. Eng. 10(21):6871-6888. https://doi.org/10.1021/acssuschemeng.2c00873
45.
Bailey JD, Helbling E, Mankar A, Stirling M, Hicks F, Leahy DK. 2021. Beyond organic solvents: synthesis of a 5-HT4 receptor agonist in water. Green Chem. 23(2):788-795. https://doi.org/10.1039/d0gc03316b
46.
Gallou F, Lipshutz BH. 2018. Organometallic Processes in Water. Springer.1-18. https://doi.org/10.1007/3418_2018_17
47.
Bihani M, Ansari TN, Finck L, Bora PP, Jasinski JB, Pavuluri B, Leahy DK, Handa S. 2020. Scalable α-Arylation of Nitriles in Aqueous Micelles using Ultrasmall Pd Nanoparticles: Surprising Formation of Carbanions in Water. ACS Catal. 10(12):6816-6821. https://doi.org/10.1021/acscatal.0c01196
48.
Ansari TN, Sharma S, Hazra S, Hicks F, Leahy DK, Handa S. 2022. Trichloromethyl Carbanion in Aqueous Micelles: Mechanistic Insights and Access to Carboxylic Acids from (Hetero)aryl Halides. ACS Catal. 12(24):15686-15695. https://doi.org/10.1021/acscatal.2c05288
49.
Brals J, Smith JD, Ibrahim F, Gallou F, Handa S. 2017. Micelle-Enabled Palladium Catalysis for Convenient sp2-sp3 Coupling of Nitroalkanes with Aryl Bromides in Water Under Mild Conditions. ACS Catal. 7(10):7245-7250. https://doi.org/10.1021/acscatal.7b02663
50.
Wood AB, Roa DE, Gallou F, Lipshutz BH. α-Arylation of (hetero)aryl ketones in aqueous surfactant media. Green Chem. 23(13):4858-4865. https://doi.org/10.1039/d1gc01572a
51.
DeAngelis AJ, Gildner PG, Chow R, Colacot TJ. 2015. Generating Active “L-Pd(0)” via Neutral or Cationic π-Allylpalladium Complexes Featuring Biaryl/Bipyrazolylphosphines: Synthetic, Mechanistic, and Structure–Activity Studies in Challenging Cross-Coupling Reactions. J. Org. Chem. 80(13):6794-6813. https://doi.org/10.1021/acs.joc.5b01005
52.
Maiti D, Fors BP, Henderson JL, Nakamura Y, Buchwald SL. 2011. Palladium-catalyzed coupling of functionalized primary and secondary amines with aryl and heteroaryl halides: two ligands suffice in most cases. Chem. Sci. 2(1):57-68. https://doi.org/10.1039/c0sc00330a
53.
Barder TE, Buchwald SL. 2007. Insights into Amine Binding to Biaryl Phosphine Palladium Oxidative Addition Complexes and Reductive Elimination from Biaryl Phosphine Arylpalladium Amido Complexes via Density Functional Theory. J. Am. Chem. Soc. 129(39):12003-12010. https://doi.org/10.1021/ja073747z
54.
Kataoka N, Shelby Q, Stambuli JP, Hartwig JF. 2002. Air Stable, Sterically Hindered Ferrocenyl Dialkylphosphines for Palladium-Catalyzed C−C, C−N, and C−O Bond-Forming Cross-Couplings. J. Org. Chem. 67(16):5553-5566. https://doi.org/10.1021/jo025732j
55.
Shelby Q, Kataoka N, Mann G, Hartwig J. 2000. Unusual in Situ Ligand Modification to Generate a Catalyst for Room Temperature Aromatic C−O Bond Formation. J. Am. Chem. Soc. 122(43):10718-10719. https://doi.org/10.1021/ja002543e
56.
Xu G, Gao P, Colacot TJ. 2022. Tunable Unsymmetrical Ferrocene Ligands Bearing a Bulky Di-1-adamantylphosphino Motif for Many Kinds of Csp2–Csp3 Couplings. ACS Catal. 12(9):5123-5135. https://doi.org/10.1021/acscatal.2c00352
57.
Su M, Buchwald SL. 2012. A Bulky Biaryl Phosphine Ligand Allows for Palladium‐Catalyzed Amidation of Five‐Membered Heterocycles as Electrophiles. Angew Chem Int Ed. 51(19):4710-4713. https://doi.org/10.1002/anie.201201244
58.
Klaus S, Neumann H, Zapf A, Strübing D, Hübner S, Almena J, Riermeier T, Groß P, Sarich M, Krahnert W, et al. 2006. A General and Efficient Method for the Formylation of Aryl and Heteroaryl Bromides. Angew Chem Int Ed. 45(1):154-158. https://doi.org/10.1002/anie.200502697
59.
Lundgren R, Peters B, Alsabeh P, Stradiotto M. 2010. A P,N‐Ligand for Palladium‐Catalyzed Ammonia Arylation: Coupling of Deactivated Aryl Chlorides, Chemoselective Arylations, and Room Temperature Reactions. Angew Chem Int Ed. 49(24):4071-4074. https://doi.org/10.1002/anie.201000526
60.
Chen L, Ren P, Carrow BP. 2016. Tri(1-adamantyl)phosphine: Expanding the Boundary of Electron-Releasing Character Available to Organophosphorus Compounds. J. Am. Chem. Soc. 138(20):6392-6395. https://doi.org/10.1021/jacs.6b03215
61.
Surry DS, Buchwald SL. 2011. Dialkylbiaryl phosphines in Pd-catalyzed amination: a user's guide. Chem. Sci. 2(1):27-50. https://doi.org/10.1039/c0sc00331j
62.
Jiang T, Kuhen KL, Wolff K, Yin H, Bieza K, Caldwell J, Bursulaya B, Wu TY, He Y. 2006. Design, synthesis and biological evaluations of novel oxindoles as HIV-1 non-nucleoside reverse transcriptase inhibitors. Part I. Bioorganic & Medicinal Chemistry Letters. 16(8):2105-2108. https://doi.org/10.1016/j.bmcl.2006.01.073
63.
Ding K, Lu Y, Nikolovska-Coleska Z, Wang G, Qiu S, Shangary S, Gao W, Qin D, Stuckey J, Krajewski K, et al. 2006. Structure-Based Design of Spiro-oxindoles as Potent, Specific Small-Molecule Inhibitors of the MDM2−p53 Interaction. J. Med. Chem. 49(12):3432-3435. https://doi.org/10.1021/jm051122a
64.
Durbin MJ, Willis MC. 2008. Palladium-Catalyzed α-Arylation of Oxindoles. Org. Lett. 10(7):1413-1415. https://doi.org/10.1021/ol800141t
65.
Altman RA, Hyde AM, Huang X, Buchwald SL. 2008. Orthogonal Pd- and Cu-Based Catalyst Systems for C- and N-Arylation of Oxindoles. J. Am. Chem. Soc. 130(29):9613-9620. https://doi.org/10.1021/ja803179s
66.
Biosca M, Jackson M, Magre M, Pàmies O, Norrby P, Diéguez M, Guiry PJ. 2018. Enantioselective Synthesis of Sterically Hindered Tertiary α‐Aryl Oxindoles via Palladium‐Catalyzed Decarboxylative Protonation. An Experimental and Theoretical Mechanistic Investigation. Adv Synth Catal. 360(16):3124-3137. https://doi.org/10.1002/adsc.201800507
67.
You J, Verkade JG. 2003. P(i-BuNCH2CH2)3N:  An Efficient Ligand for the Direct α-Arylation of Nitriles with Aryl Bromides. J. Org. Chem. 68(21):8003-8007. https://doi.org/10.1021/jo034779h
68.
Goebel JF, Löffler J, Zeng Z, Handelmann J, Hermann A, Rodstein I, Gensch T, Gessner VH, Gooßen LJ. 2023. Computer‐Driven Development of Ylide Functionalized Phosphines for Palladium‐Catalyzed Hiyama Couplings. Angew Chem Int Ed. 62(9): https://doi.org/10.1002/anie.202216160
69.
Vogl EM, Buchwald SL. 2002. Palladium-Catalyzed Monoarylation of Nitroalkanes. J. Org. Chem. 67(1):106-111. https://doi.org/10.1021/jo010953v
70.
Maryanoff CA, Sorgi KL, Zientek AM. 1994. Ketene and carbenoid behavior of .alpha.-halo ester enolates. J. Org. Chem. 59(1):237-239. https://doi.org/10.1021/jo00080a040
71.
Bartlett PA, Barstow JF. 1982. Ester-enolate Claisen rearrangement of .alpha.-amino acid derivatives. J. Org. Chem. 47(20):3933-3941. https://doi.org/10.1021/jo00141a024
72.
Agnelli F, Sulikowski GA. 1998. Synthesis of arylacetates by the palladium-catalyzed cross-coupling of aryl bromides and copper(II) enolates. Tetrahedron Letters. 39(48):8807-8810. https://doi.org/10.1016/s0040-4039(98)01978-9
73.
Giordano C, Castaldi G, Uggeri F. 1984. Synthesis of Anti‐Inflammatory α‐Arylalkanoic Acids by 1,2‐Aryl Shift. New Synthetic Methods (42). Angew. Chem. Int. Ed. Engl. 23(6):413-419. https://doi.org/10.1002/anie.198404131
74.
Akdeniz A, Mosca L, Minami T, Anzenbacher P. 2015. Sensing of enantiomeric excess in chiral carboxylic acids. Chem. Commun. 51(26):5770-5773. https://doi.org/10.1039/c5cc00376h
75.
Schoffers E, Golebiowski A, Johnson CR. 1996. Enantioselective synthesis through enzymatic asymmetrization. Tetrahedron. 52(11):3769-3826. https://doi.org/10.1016/s0040-4020(95)01021-1
76.
Grasa GA, Colacot TJ. 2007. α-Arylation of Ketones Using Highly Active, Air-Stable (DtBPF)PdX2(X = Cl, Br) Catalysts. Org. Lett. 9(26):5489-5492. https://doi.org/10.1021/ol702430a
77.
Kincaid JRA, Wong MJ, Akporji N, Gallou F, Fialho DM, Lipshutz BH. 2023. Introducing Savie: A Biodegradable Surfactant Enabling Chemo- and Biocatalysis and Related Reactions in Recyclable Water. J. Am. Chem. Soc. 145(7):4266-4278. https://doi.org/10.1021/jacs.2c13444
78.
Sharma P, Ponnusamy E, Ghorai S, Colacot TJ. 2022. DOZN™ 2.0: A quantitative green chemistry evaluator for a sustainable future. Journal of Organometallic Chemistry. 970-971122367. https://doi.org/10.1016/j.jorganchem.2022.122367