MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now
qPCR
SYBR Green-based qPCR
Quantitative Real-Time PCR (qPCR) uses fluorescent reporter molecules to allow quantification of amplified products. As in conventional PCR, a DNA, cDNA, or RNA template is amplified, but at each cycle, fluorescent signals are monitored for relative or absolute quantification.
This technique is useful for numerous areas of research including gene expression analysis, genotyping, microRNA analysis, genetic variation analysis, and protein analysis.
Featured Categories
A gloved hand holding a test tube with a green substance over a rack of other test tubes in a laboratory setting.
PCR Reagents & Kits
Polymerase Chain Reaction (PCR) is a mainstay in molecular biology labs. PCR is used for amplifying DNA by several orders of magnitude and our reagents and resources are suitable for numerous research workflows including, gene expression analysis, genotyping, sequencing, and mutagenesis application needs.
Shop Now
Molecular Cloning & Protein Expression
Molecular Cloning & Protein Expression
Molecular cloning and protein expression methods and protocols for reliable cloning and robust protein expression from insect, bacteria, and mammalian protein expression vectors.
Shop Now
Epigenetics
Epigenetics
Epigenetic reagents and resources for investigating RNA species, DNA methylation, chromatin modifications, and histone modifications.
Shop Now
Microbiome
Microbiome
Comprehensive Gut Microbiome Analysis: Uncover a holistic solution encompassing sample prep, sequencing, bioinformatics, and statistics. From 16S to WGS.
Shop Now
How to Analyze qPCR Data
The fluorescence of the reporter molecules is measured and quantified; typically these are either dyes (i.e. SYBR® Green, Ethidium Bromide) that intercalate between the bases in double-stranded DNA, or probes designed to bind a specific sequence (i.e. Molecular Beacons, TaqMan® probes) on the DNA.
Relative Quantitation of qPCR Data
There are two major quantification methods for qPCR data. Relative quantification, the more common method, uses ΔΔCt information, by which the expression or abundance ratio of the target gene in the sample is determined, compared to a control gene, and normalized with the expression ratio of a reference gene. Since, the efficiency E values for target and reference genes differ, this method also accounts for the differences in E values. This method is more frequently employed in gene expression analysis.
Absolute Quantitation of qPCR Data
The second method, absolute quantification, is more common in environmental microbiology and makes use of the standard-curve (SC). The SC method uses dilution series of known template concentration, N0, to create a standard curve using the linear regression of log(N0) versus the CT (threshold cycle). This is then used to calculate template concentrations of the sample. The basis of this method is that the efficiency value of the sample is the same as that of the standard. 
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents