Understanding Lab Water: The Foundation of Precision Science
Water is one of the most used reagents in laboratories — and one of the least understood. Whether you're in pharmaceuticals, diagnostics, the food and beverage industry, or a biotech laboratory, water quality directly impacts results, compliance, and operations. This hub explores what water purity means, why it matters, and how to optimize its purity for your specific scientific analyses.
Featured Categories
Milli-Q® systems offer innovative water purification technologies engineered to support lab research needs, sustainability goals, and other major requirements.
From a single lab to an entire building, Milli-Q® central water purification systems are connected, high-performance instruments that can reliably feed all purified water needs.
Find the cartridges, UV lamps, filters and polishers for your water system. Advantages of using genuine Milli-Q consumables: guarantee water quality, sustainability features (new Drain Cap, Hg-free lamps). Designed for perfect fit.
Milli-Q® water system accessories support lab-specific needs, improving safety, water quality, and productivity while conserving bench space.
The critical role of water in the lab
As a universal solvent, water is used in nearly every laboratory reagent and application, whether for rinsing, buffer or sample dilution, or as a blank. The quality of water used in the lab — and its consistency of quality — is therefore critical for the success of the tests performed.
Properties of Water
It’s important to understand why water is such a unique molecule. Read the article Water: Properties, Polarity and Its Role as a Universal Solvent to learn about its polarity, hydrogen bonding, and how these properties make it a universal solvent essential for life and work in the laboratory.
Water Contaminants
Water impurities are generally classified into five major classes: organics, inorganic ions, bacteria and their by-products, particles and colloids, and dissolved gas. Learn the sources of these water contaminants, their impacts on various laboratory instruments and experiments, and how to effectively remove them from your water supply in the article, Water Contaminants: Types, Sources and Impacts in the Lab.
Water Purification Techniques
There are many different water purification techniques and technologies to remove contaminants from lab water. These include distillation, reverse osmosis, ion exchange resins, ultrafilters, carbon filtration and UV oxidation. Each technology has benefits and limitations. Some can remove a large fraction of several contaminants (e.g., reverse osmosis), while others excel at removing one specific type of impurity down to very low levels (e.g. ion exchange resins). As no single technology can remove all contaminants, several purification technologies are generally combined to produce pure and ultrapure water.
Water Quality Grades
Norms define different laboratory water grades for both technical and economical reasons. The purpose of these norms is to ensure that the right water quality is used for a specific application, while limiting laboratory operating costs as Type 1 ultrapure water is more expensive to produce than Type 2 or Type 3 pure water.
Determine which water grade is best suited for your specific analysis, instrument or equipment in the article, Understanding Water Quality Grades for Laboratory Applications. Ensuring that you use the optimal water type for your application will support results reliability and accuracy, while also optimizing laboratory costs.
Monitoring of Water Quality
Ensuring the purity of water used in laboratory experiments is crucial for obtaining accurate and reliable results. Contaminants such as inorganic salts and dissolved organics can significantly impact the integrity of scientific research. The article Water Quality Monitoring describes the importance of monitoring water quality in the lab, highlighting advanced techniques for detecting ionic and organic impurities via online resistivity and TOC measurements.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Choosing a water purification system? Consider required water quality, total volume, monitoring & traceability, and service needs.
- This study illustrates the efficiency of Milli-Q® water purification systems in eliminating pesticides from tap water for reliable LC-MS results.
- Best practices for microbial testing of purified water in the lab. Covers storage, culture and incubation conditions, and contaminants reduction.
- Explore how various water purification technologies effectively remove contaminants to ensure laboratory-grade water quality for scientific analyses.
- Learn about various water contaminants, their sources, impacts on laboratory experiments, and effective removal methods.
- See All (11)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email custserv@sial.com
or call +1 (800) 244-1173
Additional Support
- Lab Water Solutions Support
Receive professional assistance in identifying the optimal water solution for your laboratory.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ
Explore our Frequently Asked Questions for answers to commonly asked questions about our products and services.