Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

P5-48-XT-A

Air Science USA LLC

Air Science Ductless Fume Hood

Ductless Fume Hood 48" Wide

Shipped & Sold By Air Science

A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.

Select a Size


Marketplace Support
Need help or have questions about this product?
Contact Seller
Marketplace Support
Need help or have questions about this product?
Contact Seller

General description

Ductless Fume Hood 48" Wide Basic

Purair®BASIC Ductless Fume Hood, 48" / 1200mm Nominal Width, Comprising of a P5-48-HEAD-A and a P5-48-XT-ENCL, Standard Depth, 115V 60Hz, North American Cord set (unless specified). Requires Minimum Two ASTS-XXX Main Filters installed side-by-side (various types based on your chemical handlings) sold separately: ASTS-001 GP Plus! filter for solvents, ASTS-015 ACI Plus! filter for acids, ASTS-012 AMM Filter for ammonia, ASTS-013 FOR Filter for formaldehydes, and ASTS-030 HEPA filter for particulates


Purair Basic ductless fume hoods are designed to provide high-level performance features at an affordable price. The Purair Basic is designed to protect the user and the environment from hazardous vapors generated on the work surface. Compact basic series ductless fume hoods are ideal for areas with limited space and applications that generate small volumes of dangerous or potentially dangerous vapors. The 48" Purair Basic ductless fume hood is found in research laboratories, educational facilities, and forensic laboratories throughout the world.


Specifications:

  • Controller Type: Standard Control Panel
  • Voltage: 115V 60Hz
  • Nominal Width (feet): 4ft
  • Depth: Standard Depth
  • Height: Extra Tall Height
  • Pack Size: Each
  • UOM: EA

Design Features:

  • Filter I.D. Window: A convenient, strategically placed front cover window shows the installed filter part number and installation date to encourage timely filter replacement.
  • Control Panel: Electronic controls and displays include switches for the blower and filter blockage alarm.
  • Filter Blockage Alarm: Continuously monitors filter loading and alerts user when service is needed.
  • Optional Air Velometer: An optional analog air velocity meter is positioned in the user's field of vision.
  • Steel Support Frame: The chemical resistant epoxy coated steel frame adds mechanical strength.
  • Hinged Front Sash: When closed, the cabinet sash protects the contents from inadvertent external contact and better isolates the air within. The sash is easy to open and close.
  • Optional Work Surface: The internal work surface can be fitted with an optional polypropylene (available in white and black) or stainless steel tray.
  • Pass Through Ports: Electrical cords and cables are safely routed into the cabinet through ports an the back.
  • Electrostatic Pre-Filter: The electrostatic pre-filter is accessible from inside the chamber and 91% effective down to 1-3 microns.
  • Filter Door Key: Filter access keys prevent unauthorized removal or accidental exposure to dirty filters.
  • Internal Manual Speed controller: Authorized personnel may adjust blower speed as desired.
  • Optional Base Stand: Optional mobile cart with locking casters.
  • Task Lighting: LED task light illuminates the workzone.
  • Additional Features:

    • 360 Degree Visibility: Clear back and side panels allow ambient light into the chamber and provide users with an unobstructed view of contents.

    Options and Accessories Sold Separately:

    • Base Stand: CART-50 Mobile Base Stand.
    • Spill Tray: TRAY-48 Black Polypropylene Removeable Work Surface with Spill Lip .
    • DWYER Airflow Meter: DWYER Airflow Meter Continuously monitors airflow.

Legal Information

Purair is a registered trademark of Air Science USA LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library