MilliporeSigma
ALPHA-321-02-701
AlphaGem Bio, Inc
AlphaGem qPCR Tube
.2ml 8 strip tubes and optical strip caps/ co molded RT/Qpcr caps
Shipped & Sold By AlphaGem Bio, Inc
A Trusted Marketplace Seller
Synonym(s):
PCR Tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
material
polypropylene
manufacturer/tradename
AlphaGem Bio, Inc
General description
Introducing our high-quality PCR strips and tubes made from medical-grade plastic, to ensure their safety in sensitive applications. Our PCR strips and tubes are equipped with a unique patented lens design, which improves sensitivity and accuracy in detecting and measuring the amount of amplified DNA, along with ergonomic caps that significantly reduce installation force by over 10X and improve workplace ergonomics without sample evaporation. With uniform thin walls for optimal heat transfer and low evaporation rates during thermal cycling, our PCR strips and tubes offer consistent, reliable results. Available in various sizes and styles, including single tubes, 8-strip tubes, and ergonomic 8-strip tubes, our PCR strips and tubes are perfect for any experiment needs. With their excellent features and quality, our PCR strips and tubes are the perfect choices for lab professionals, researchers, and scientists looking for reliable, accurate results every time.
Still not finding the right product?
Explore all of our products under AlphaGem qPCR Tube
Choose from one of the most recent versions:
Certificates of Analysis (COA)