Sélectionner une taille de conditionnement
binding type
non-treated surface
feature
graduations: Yes
sterility
gamma-irradiated
volume
200 μL
manufacturer/tradename
AlphaGem Bio, Inc
General description
Premium 96-format automation pipette tips that can transform your liquid handling processes. These tips are manufactured with high-quality medical-grade polypropylene, which provides optimal chemical resistance and compatibility with different reagents. With universal design and liquid holding capacities, our automation tips are compatible with the most popular brands and models of automated liquid handlers making them a versatile choice for any laboratory.
Whether you work in genomics, drug discovery, or clinical diagnostics, our automation pipette tips will improve your workflow by providing accurate and consistent liquid transfers every time.
96 Tips Per Rack. 10 Racks Per Unit. 5 Units Per Case.
Faites votre choix parmi les versions les plus récentes :
Déjà en possession de ce produit ?
Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.