Skip to Content
MilliporeSigma

EF-4-38-SYS

ECO Funnel®

CP Lab Safety 4 inch ECO Funnel® Complete System

Includes 4 Liter LDPE Bottle, 38mm ECO Funnel, Secondary Container, Red Lid

Shipped & Sold By CP Lab Safety

A Trusted Marketplace Seller

Synonym(s):

safety funnel, funnel with lid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


capacity

450 mL

L

13 in

material

PE

top diam.

5 in

manufacturer/tradename

CP Lab Safety

General description

This 4" ECO Funnel® system includes a 4 liter LDPE bottle with a 38-430 closure and a EPA compliant secondary container with detachable base.

  • Dimensions: Lid Open: 13"L x 13"W x 23.5"H
  • Dimensions: Lid Closed: 13L x 13"W x 18"H
  • Funnel: 4" ECO Funnel® EF-4-38
  • Container: 4 Liter LDPE EC-2004
  • Secondary Container: SC-3004CPB

Specifications:

  • 4" diameter opening
  • 450 mL fluid capacity
  • Latching lid with inner gasket
  • Chemical resistant HDPE construction
  • Removable stir-bar catcher screen
  • Detachable tube reduces chemical evaporation of hazardous solvents
  • Can be custom fitted with up to 6 HPLC waste lines
  • Funnel top without the stem measures 5" W x 7"L
  • Made in the USA

Benefits of ECO Funnel®

  • Solves the "open waste container" problem.
  • Prevents emissions of volatile waste solvents, acids, and bases in the laboratory.
  • Has a unique, patented design that meets most local, state and federal safety regulations and requirements.
  • May be used again and again and resists nearly all corrosive chemicals.
  • Is considered a state of the art product by environmental health and safety officers and industry professionals.
  • Applies to Pharmaceutical, Biotech, Academic and Government laboratories and is widely used.
  • Small footprint allows for use in a fume hood

The ECO Funnel is the ultimate lab safety product for collection of hazardous liquid waste. Its unique, patented design performs two functions, acting as both a sealed waste container and also as a solvent funnel. The 4" ECO Funnel® is made of HDPE and has a 450 mL fluid capacity. This safety funnel is designed with a custom screw cap which secures it to a specific waste container. It features a latching lid with gasket to keep fumes contained and a removable large particle filter catches stir bars or other items. The detachable tube extends into the waste container to prevent chemical evaporation. The ECO Funnel is a pass-through system, meaning chemicals are not sitting in the funnel for any length of time, so it's HDPE construction is compatible with most solvents including those that need to be stored in glass.

ECO Funnel systems are available in various sizes with and without EPA compliant secondary containment, which is designed to hold 10% more than the primary container and safety contain spills and containment failures. Read about secondary containment regulations and compliance.

Legal Information

ECO Funnel is a registered trademark of CP Lab Safety


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library