MilliporeSigma
EISCO-BI0139
Eisco Labs
Eisco Labs 8 Piece Plastic Sieve/ Strainer
8 Piece Plastic Sieve/ Strainer
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
Synonym(s):
Sieve, Strain, Sift, Filter
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Eisco Labs
General description
  • Six plastic sieves with stainless steel mesh, cover and Bottom Collector.
  • Each Sieve has a plastic frame having an individual size one each ( Mesh no. 5, 10, 35, 60, 120, 230)
  • Complete with plastic lid and bottom collector
  • Stackable - Diameter of Sieves are 6.75"
  • Total height of stacked sieves approx. 15"
Eisco Labs 8 Piece Plastic Sieve/ Strainer - Stainless Steel Mesh Six plastic sieves with stainless steel mesh. Each Sieve has a plastic frame having an individual size one each ( Mesh no. 5, 10, 35, 60, 120, 230) Complete with plastic lid and bottom collector. Stackable, Total height of stacked sieves approx. 15.25" Not not food.
Choose from one of the most recent versions:
Certificates of Analysis (COA)